Skip to Content
Merck
All Photos(1)

Key Documents

SML4022

Sigma-Aldrich

Didemnin B

new

≥95% (HPLC)

Synonym(s):

(2S)-2-Hydroxypropanoyl-L-prolyl-N-methyl-D-leucyl-L-threonyl-(3S,4R,5S)-4-amino-3-hydroxy-5-methylheptanoyl-(2S,4S)-4-hydroxy-2,5-dimethyl-3-oxohexanoyl-L-leucyl-L-prolyl-N,O-dimethyl-L-tyrosine (9→4)-lactone, N-[1-[(2S)-2-Hydroxy-1-oxopropyl]-L-prolyl]didemnin A, NSC 325319, NSC 333841

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C57H89N7O15
CAS Number:
Molecular Weight:
1112.35
UNSPSC Code:
12352209
UNSPSC Code:
12352200

Quality Level

Assay

≥95% (HPLC)

form

(Powder or Lyophilized powder or film)

color

white to off-white

storage temp.

-10 to -25°C

SMILES string

O=C1N2[C@]([H])(C(N(C)[C@@H](CC3=CC=C(OC)C=C3)C(O[C@H](C)[C@H](NC([C@H](N(C([C@H]4N(C([C@H](C)O)=O)CCC4)=O)C)CC(C)C)=O)C(N[C@]([H])([C@H](CC)C)[C@@H](O)CC(O[C@@H](C(C)C)C([C@H](C)C(N[C@H]1CC(C)C)=O)=O)=O)=O)=O)=O)CCC2

InChI

1S/C57H89N7O15/c1-15-33(8)46-44(66)29-45(67)79-49(32(6)7)48(68)34(9)50(69)58-39(26-30(2)3)54(73)64-25-17-19-41(64)56(75)62(13)43(28-37-20-22-38(77-14)23-21-37)57(76)78-36(11)47(52(71)59-46)60-51(70)42(27-31(4)5)61(12)55(74)40-18-16-24-63(40)53(72)35(10)65

InChI key

KYHUYMLIVQFXRI-SJPGYWQQSA-N

Biochem/physiol Actions

Marine-derived cyclic depsipeptide with potent antiviral, immunosuppressive and anticancer activities; induces rapid apoptosis; elongation factor 1A inhibitor.Didemnin B is a marine-derived cyclic depsipeptide with potent antiviral, immunosuppressive and anticancer activities. Didemnin B selectively induces rapid apoptosis through dual inhibition of palmitoyl-protein thioesterase 1 (PPT1) and eukaryotic translation elongation factor 1 alpha 1 (EEF1A1). Binds to the eEF1A(GTP)-aa-tRNA ternary complex and inhibits translation. Didemnin B improves nonalcoholic fatty liver disease (NAFLD) histopathology, glucose homeostasis, and dyslipidemia in western diet-induced obese mice.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Malia B Potts et al.
Nature chemical biology, 11(6), 401-408 (2015-04-14)
Modern cancer treatment employs many effective chemotherapeutic agents originally discovered from natural sources. The cyclic depsipeptide didemnin B has demonstrated impressive anticancer activity in preclinical models. Clinical use has been approved but is limited by sparse patient responses combined with
Treatment with didemnin B, an elongation factor 1A inhibitor, improves hepatic lipotoxicity in obese mice
Physiological Reports, 4(17) (2016)
Manuel F Juette et al.
eLife, 11 (2022-10-21)
Rapid and accurate mRNA translation requires efficient codon-dependent delivery of the correct aminoacyl-tRNA (aa-tRNA) to the ribosomal A site. In mammals, this fidelity-determining reaction is facilitated by the GTPase elongation factor-1 alpha (eEF1A), which escorts aa-tRNA as an eEF1A(GTP)-aa-tRNA ternary

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service