Skip to Content
Merck
All Photos(2)

Key Documents

SML3800

Sigma-Aldrich

Selumetinib

≥98% (HPLC)

Synonym(s):

5-((4-Bromo-2-chlorophenyl)amino)-4-fluoro-N-(2-hydroxyethoxy)-1-methyl-1H-benzo[d]imidazole-6-carboxamide, 6-(4-Bromo-2-chloro-phenylamino)-7-fluoro-3-methyl-3H-benzoimidazole-5-carboxylic acid (2-hydroxy-ethoxy)-amide, ARRY 142886, ARRY-142886, ARRY142886, AZD 6244, AZD-6244, AZD6244

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H15BrClFN4O3
CAS Number:
Molecular Weight:
457.68
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.21

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

2-8°C

Biochem/physiol Actions

Selumetinib (ARRY-142886; AZD6244) is an orally active, potent and selective MEK1/2 inhibitor (MEK1 IC50 = 14 nM; <20% inhibition of >40 other kinases at 10 µM) that effectively downregulates cellular ERK1/2 phosphorylation levels (IC50 <40 nM post 1h incubation using 5 different cell lines). Selumetinib inhibits MEK1/2-dependent proliferations in cancer cultures (IC50 in nM = 59/Malme-3M, 93/SK-MEL-28, 175/HT-29, 200/SK-MEL-2, 473/MIA PaCa-2) and exhibits anti-tumor efficacy in mice in vivo (10-100 mg/kg p.o. b.i.d.; BxPC3 & HT-29 xenograft models).

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 4 - Repr. 2 - Skin Sens. 1 - STOT RE 2

Target Organs

Gastrointestinal tract,Eyes,Urinary tract

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Barry R Davies et al.
Molecular cancer therapeutics, 6(8), 2209-2219 (2007-08-19)
Constitutive activation of the extracellular signal-regulated kinase 1/2 (ERK1/2) mitogen-activated protein kinase (MAPK) signaling pathway in human cancers is often associated with mutational activation of BRAF or RAS. MAPK/ERK kinase 1/2 kinases lie downstream of RAS and BRAF and are
Tammie C Yeh et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 13(5), 1576-1583 (2007-03-03)
The Ras-Raf-mitogen-activated protein kinase kinase (MEK) pathway is overactive in many human cancers and is thus a target for novel therapeutics. We have developed a highly potent and selective inhibitor of MEK1/2. The purpose of these studies has been to
Nikolas K Haass et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 14(1), 230-239 (2008-01-04)
Disseminated melanoma is highly therapy resistant. The finding that 66% of melanomas harbor the activating BRAF(V600E) mutation has raised expectations for targeting the Ras/RAF/mitogen-activated protein (MAP)/extracellular signal-regulated kinase (ERK) kinase (MEK)/ERK pathway in melanoma. This study addresses the anti-melanoma activity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service