Skip to Content
Merck
All Photos(1)

Key Documents

SML3274

Sigma-Aldrich

AZD5904

≥95% (HPLC)

Synonym(s):

(R)-3-(2-Tetrahydrofuryl-methyl)-2-thioxanthine, (R)-TX3, 3-[[(2R)-Tetrahydrofuran-2-yl]methyl]-2-thioxo-7H-purin-6-one, AZD 5904, AZD-5904, TX3 R-enantiomer, TX4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12N4O2S
CAS Number:
Molecular Weight:
252.29
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

O=C1NC(N(C[C@@H]2OCCC2)C3=C1NC=N3)=S

InChI

1S/C10H12N4O2S/c15-9-7-8(12-5-11-7)14(10(17)13-9)4-6-2-1-3-16-6/h5-6H,1-4H2,(H,11,12)(H,13,15,17)/t6-/m1/s1

InChI key

RSPDBEVKURKEII-ZCFIWIBFSA-N

Biochem/physiol Actions

AZD5904 (TX4) is an orally active 2-thioxanthine class suicide substrate that targets myeloperoxidase (MPO) for mechanism-based inactivation, covalently modifying MPO heme group without converting the enzyme to compound II. AZD5904 effectively inhibits peroxide-induced human MPO chlorination activity (IC50 = 0.2 μM) and extracellular MPO activity in PMA-stimulated human neutrophil cultures (by 68% at 1 μM). Oral administration (20-180 μmol/kg) of the racemate (TX3) is efficacious in reducing inflammation site MPO activity in mice in vivo with 10- to 19-fold selectivity over lactoperoxidase (LPO), thyroid peroxidase (TPO), and >70-fold selectivity over a panel of other enzymes, ion channels, and receptors.
Highly selective and orally available mechanism-based myeloperoxidase (MPO) inactivatior with in vitro and in vivo efficacy.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chrishan J A Ramachandra et al.
Cardiovascular research, 118(2), 517-530 (2021-03-12)
Hypertrophic cardiomyopathy (HCM) is characterized by cardiomyocyte hypertrophy and disarray, and myocardial stiffness due to interstitial fibrosis, which result in impaired left ventricular filling and diastolic dysfunction. The latter manifests as exercise intolerance, angina, and dyspnoea. There is currently no
Reena J Popat et al.
JCI insight, 2(2), e87379-e87379 (2017-02-01)
Anti-neutrophil cytoplasmic antibody (ANCA) vasculitis is characterized by the presence of autoantibodies to myeloperoxidase and proteinase-3, which bind monocytes in addition to neutrophils. While a pathological effect on neutrophils is acknowledged, the impact of ANCA on monocyte function is less
Weidong Chai et al.
American journal of physiology. Endocrinology and metabolism, 317(6), E1063-E1069 (2019-10-09)
A high-fat diet (HFD) can rapidly recruit neutrophils to insulin target tissues and within days induce microvascular insulin resistance (IR). Myeloperoxidase (MPO) is highly enriched in neutrophils, can inhibit nitric oxide-mediated vasorelaxation in vitro and is associated with increased cardiovascular
Lars B Nilsson et al.
Rapid communications in mass spectrometry : RCM, 26(12), 1399-1406 (2012-05-18)
The investigations in this article were triggered by two observations in the laboratory; for some liquid chromatography/tandem mass spectrometry (LC/MS/MS) systems it was possible to obtain linear calibration curves for extreme concentration ranges and for some systems seemingly linear calibration
Halla Björnsdottir et al.
Free radical biology & medicine, 89, 1024-1035 (2015-10-16)
Neutrophil extracellular traps (NETs) are mesh-like DNA fibers clad with intracellular proteins that are cast out from neutrophils in response to certain stimuli. The process is thought to depend on reactive oxygen species (ROS) generated by the phagocyte NADPH-oxidase and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service