Skip to Content
Merck
All Photos(1)

Documents

SML3140

Sigma-Aldrich

GsMTx4 trifluoroacetate

≥85% (HPLC), powder, TRPC1 and TRPC6 inhibitor

Synonym(s):

1-amino-Cy5-glucose, Cyanine-based 1-amino-1-deoxy-β-glucose conjugate, Cy5-linked 1amino-glucose, GCLEFWWKCNPNDDKCCRPKLKCSKLFKLCNFSF-NH2 trifluoroacetate(Disulfide: 2-17, 9-23, 16-30), Gly-Cys-Leu-Glu-Phe-Trp-Trp-Lys-Cys-Asn-Pro-Asn-Asp-Asp-Lys-Cys-Cys-Arg-Pro-Lys-Leu-Lys-Cys-Ser-Lys-Leu-Phe-Lys-Leu-Cys-Asn-Phe-Ser-Phe-NH2 trifluoroacetate (Disulfide: 2-17, 9-23, 16-30), Grammostola Mechanotoxin #4 trifluoroacetate, GsMTx-4 trifluoroacetate, L-GsMTx trifluoroacetate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.77

product name

GsMTx4 trifluoroacetate, ≥85% (HPLC)

Quality Level

Assay

≥85% (HPLC)

form

powder

color

white to beige

storage temp.

−20°C

General description

Grammostola mechanotoxin #4 (GsMTx4), a gating modifier peptide, is obtained from spider venom. This 34-amino-acid peptide is a small (3-5 kD) amphipathic molecule built on a conserved inhibitory cysteine-knot (ICK) backbone. The structure of GsMTx4 has a belt of charged residues surrounding a hydrophobic patch. GsMTx4 acts as an inhibitor of cationic mechanosensitive channels (MSCs) belonging to the Piezoand transient receptor potential (TRP) channel families. It is different from other spider venom peptides because it can inhibit these channels regardless of their shape, and both its L- and D-forms can have this effect. GsMTx4 has a unique structure and carries a positive charge due to its six lysine residues which can affect membrane binding. This peptide has become a valuable tool in research to identify the roles of excitatory MSCs in normal physiology and pathology. It can also inhibit stretch-activated big potassium channels (SAKcaC). The presence of positively charged residues in GsMTx4 is thought to play a role in peptide channel-lipid interaction. It can effectively reduce arrhythmias by inhibiting stretch-activated channels (SACs) in the heart. GsMTx4 also serves as an inhibitor of histone acetylation inhibitor I-BET 762 and Piezo 1 inhibitor.

Application

GsMTx4 trifluoroacetate has been used in drug treatment as a supplement in cell culture medium to treat mouse hippocampal neurons to block cationic MSCs in cortical neurons. It has also been used as a specific inhibitor to incubate cementoblasts to study its effects on the response of cementoblasts to investigate if cementum repair was impaired by compression.

Biochem/physiol Actions

GsMTx4 (Grammostola Mechanotoxin #4) is a tarantula venom-derived 34-mer amphipathic cysteine knot peptide that exerts its cationic mechanosensitive channels (MSCs; stretch-gated ion channels or SACs) modulatory activity by changing local membrane properties near the channels. GsMTx4 negatively regulates TRPC1/5/6 and Piexo1/2, while potentiating TREK-1 and TRPA1 activity. In addition, GsMTx4 can either hamper (2-4 μM) or facilitate (12-20 μM) the gating of prokaryotic E. coli cytoplasmic membrane MscS and MscK in a biphasic manner. GsMTx4 is also reported to inhibit Nav1.1 (SCN1A), Nav1.2 (SCN2A), Nav1.3 (SCN3A), Nav1.4 (SCN4A), Nav1.5 (SCN5A), Nav1.6 (SCN8A), Nav1.7 (SCN9A), hERG1/2/3 (KCNH2/6/7), but not hKV1.1 (KCNA1) or hKV1.4 (KCNA4).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Fabian et al.
Pflugers Archiv : European journal of physiology, 457(2), 475-484 (2008-06-11)
Cell migration depends on the generation of structural asymmetry and on different steps: protrusion and adhesion at the front and traction and detachment at the rear part of the cell. The activity of Ca(2+) channels coordinate these steps by arranging
T M Suchyna et al.
The Journal of general physiology, 115(5), 583-598 (2000-04-25)
We have identified a 35 amino acid peptide toxin of the inhibitor cysteine knot family that blocks cationic stretch-activated ion channels. The toxin, denoted GsMTx-4, was isolated from the venom of the spider Grammostola spatulata and has <50% homology to
Annette C Hurst et al.
European biophysics journal : EBJ, 38(4), 415-425 (2009-01-10)
The spider peptide GsMTx4, at saturating concentration of 5 muM, is an effective and specific inhibitor for stretch-activated mechanosensitive (MS) channels found in a variety of eukaryotic cells. Although the structure of the peptide has been solved, the mode of
GsMTx4: Mechanism of Inhibiting Mechanosensitive Ion Channels
Gnanasambandam R, et al.
Biophysical Journal, 31-45 (2017)
Kerstin Hill et al.
The Journal of biological chemistry, 282(10), 7145-7153 (2007-01-16)
TRPA1, a poorly selective Ca(2+)-permeable cation channel, is expressed in peripheral sensory neurons, where it is considered to contribute to a variety of sensory processes such as the detection of painful stimuli. Furthermore, TRPA1 was also identified in hair cells

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service