Skip to Content
Merck
All Photos(1)

Key Documents

SML3013

Sigma-Aldrich

LY2886721

≥98% (HPLC)

Synonym(s):

LY 2886721, LY-2886721, N-(3-((4aS,7aS)-2-Amino-4a,5,7,7a-tetrahydro-4H-furo[3,4-d][1,3]thiazin-7a-yl)-4-fluorophenyl)-5-fluoropicolinamide, N-[3-[(4aS ,7aS )-2-Amino-4a,5-dihydro-4H-furo[3,4-d ][1,3]thiazin-7a(7H )-yl]-4-fluorophenyl]-5-fluoro-2-pyridinecarboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H16F2N4O2S
CAS Number:
Molecular Weight:
390.41
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

FC1=CC=C(NC(C2=CC=C(C=N2)F)=O)C=C1[C@@]34[C@@](CSC(N)=N4)([H])COC3

InChI

1S/C18H16F2N4O2S/c19-11-1-4-15(22-6-11)16(25)23-12-2-3-14(20)13(5-12)18-9-26-7-10(18)8-27-17(21)24-18/h1-6,10H,7-9H2,(H2,21,24)(H,23,25)/t10-,18-/m0/s1

InChI key

NIDRNVHMMDAAIK-YPMLDQLKSA-N

Biochem/physiol Actions

Y2886721 is an orally active, potent and selective β-secretase (BACE) active site inhibitor (human BACE1/2 IC50 = 20.3/10.2 nM; cathepsin D/pepsin/renin IC50 >10 μM). Y2886721 exhibits amyloid β-lowering efficacy in cultures (Aβ1-40/Aβ1-42 IC50 = 18.5/19.7 nM with APP751 Swedish mutation-expressing HEK293 and 10.7/9.2 nM with primary cortical neurons from PDAPP transgenic mouse embryos) and in animal models in vivo (3-30 mg/kg PDAPP mice, 1.5 mg/kg beagle dogs; p.o.).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Patrick C May et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 35(3), 1199-1210 (2015-01-23)
BACE1 is a key protease controlling the formation of amyloid β, a peptide hypothesized to play a significant role in the pathogenesis of Alzheimer's disease (AD). Therefore, the development of potent and selective inhibitors of BACE1 has been a focus
Tugce Munise Satir et al.
Alzheimer's research & therapy, 12(1), 63-63 (2020-05-28)
Alzheimer's disease (AD) is the most common form of age-related neurodegenerative diseases. Cerebral deposition of Aβ peptides, especially Aβ42, is considered the major neuropathological hallmark of AD and the putative cause of AD-related neurotoxicity. Aβ peptides are produced by sequential
Akihiro Takano et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 60(7), 992-997 (2018-12-12)
β-secretase 1 (BACE1) is a key enzyme in the generation of β-amyloid, which is accumulated in the brain of Alzheimer disease patients. PF-06684511 was identified as a candidate PET ligand for imaging BACE1 in the brain and showed high specific
Jan Schejbal et al.
Journal of separation science, 42(5), 1067-1076 (2019-01-22)
Capillary electrophoresis integrated immobilized enzyme reactors are becoming an increasingly popular alternative for enzyme kinetic and inhibition assays thanks to their unique set of features including cost effectiveness, repeated use of the enzyme, minuscule sample consumption, rapid analysis time and
Doaa M Hanafy et al.
Nutrients, 12(5) (2020-05-14)
Alzheimer's disease (AD) is a progressive neurodegenerative disorder with an unclear cause. It appears that multiple factors participate in the process of neuronal damage including oxidative stress and accumulation of the protein amyloid β (Aβ) in the brain. The search

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service