Skip to Content
Merck
All Photos(1)

Key Documents

SML2953

Sigma-Aldrich

MGR1

≥98% (HPLC)

Synonym(s):

2,2-Dimethylpropanoic acid [(1,4,4a,9,9a,10-hexahydro-9,10-dioxo-1,4-ethanoanthracen-5-yl)oxy]methyl ester, JCHD precursor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H24O5
CAS Number:
Molecular Weight:
368.42
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C1C2C(C(C3=C1C=CC=C3OCOC(C(C)(C)C)=O)=O)C4C=CC2CC4

Biochem/physiol Actions

Effective reactive oxygen species (ROS) generator in live cells. Activated by esterases to form JCHD for superoxide generation from molecular oxygen.
MGR1 is an effective reactive oxygen species (ROS) generator in live cells (dosing range 2-100 μM). MGR1 is activated by intracellular esterases to form JCHD (5-hydroxy-1,4,4a,9a-tetrahydro-1,4-ethanoanthracene-9,10-dione), which in turn, facilitated by a proximal H-bonding hydroxyl group, undergoes keto-enol tautomerism for enhanced superoxide (O2•-) generation upon reacting with molecular oxygen (O2). MGR1, but not its inactive analog MGR2, upregulates cellular ROS, resulting in cell death when used at high concentrations (>5 μM; RAW264.7, THP1, HeLa, MCF7, A549).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service