Skip to Content
Merck
All Photos(1)

Key Documents

SML2894

Sigma-Aldrich

HPA-12

≥98% (HPLC)

Synonym(s):

(1R,3S)-HPA-12, (1R,3S)-N-(3-Hydroxy-1-hydroxymethyl-3-phenylpropyl)dodecanamide, 16ASC1859, N-[(1R,3S)-3-Hydroxy-1-(hydroxymethyl)-3-phenylpropyl]dodecanamide, N-[(2R,4S)-1,4-Dihydroxy-4-phenylbutan-2-yl]dodecanamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H37NO3
CAS Number:
Molecular Weight:
363.53
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

N([C@@H](CO)C[C@H](O)c1ccccc1)C(=O)CCCCCCCCCCC

InChI

1S/C22H37NO3/c1-2-3-4-5-6-7-8-9-13-16-22(26)23-20(18-24)17-21(25)19-14-11-10-12-15-19/h10-12,14-15,20-21,24-25H,2-9,13,16-18H2,1H3,(H,23,26)/t20-,21+/m1/s1

InChI key

YCAKBKAOFSILDC-RTWAWAEBSA-N

Biochem/physiol Actions

(1R,3S)-HPA-12 is a ceramide (Cer) analog that acts as a Cer transporter (CERT) antagonist and selectively inhibits cellular Cer conversion to sphingomyelin (SM), but not to glucosylceramide, by blocking CERT-mediated Cer ER-to-Golgi transport. Common culture dosing range: 1-10 μM.
Ceramide transporter (CERT) antagonist that inhibits Cer-to-sphingomyelin (SM) conversion by blocking CERT-mediated ceramide ER-to-Golgi transport.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Masaharu Ueno et al.
Organic letters, 15(11), 2869-2871 (2013-05-25)
In response to Berkeš's report revising the stereochemistry of HPA-12, an important ceramide-trafficking inhibitor that was discovered and synthesized and its stereochemistry determined in 2001, the synthesis and the stereochemistry were reinvestigated. A large-scale synthetic method for HPA-12 based on
Hannah Scheiblich et al.
Journal of neurochemistry, 143(5), 534-550 (2017-09-25)
Inflammation within the CNS is a major component of many neurodegenerative diseases. A characteristic feature is the generation of microglia-derived factors that play an essential role in the immune response. IL-1β is a pro-inflammatory cytokine released by activated microglia, able
Noriyuki Otsuki et al.
Journal of virology, 92(1) (2017-10-27)
Rubella virus (RuV) causes a systemic infection, and transplacental fetal infection causes congenital rubella syndrome. In this study, we showed that treatment of cells with sphingomyelinase inhibited RuV infection. Assays using inhibitors of serine palmitoyl transferase and ceramide transport protein
Cécile L Bandet et al.
Diabetes, 67(7), 1258-1271 (2018-05-16)
One main mechanism of insulin resistance (IR), a key feature of type 2 diabetes, is the accumulation of saturated fatty acids (FAs) in the muscles of obese patients with type 2 diabetes. Understanding the mechanism that underlies lipid-induced IR is
S Yasuda et al.
The Journal of biological chemistry, 276(47), 43994-44002 (2001-09-08)
Ceramide produced at the endoplasmic reticulum (ER) is transported to the lumen of the Golgi apparatus for conversion to sphingomyelin (SM). N-(3-Hydroxy-1-hydroxymethyl-3-phenylpropyl)dodecanamide (HPA-12) is a novel analog of ceramide. Metabolic labeling experiments showed that HPA-12 inhibits conversion of ceramide to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service