Skip to Content
Merck
All Photos(1)

Key Documents

SML2633

Sigma-Aldrich

Sorafenib tosylate

≥98% (HPLC)

Synonym(s):

4-[4-[[[[4-Chloro-3-(trifluoromethyl)phenyl]amino]carbonyl]amino]phenoxy]-N-methyl-2-pyridinecarboxamide, 4-methylbenzenesulfonate, BAY 43-9006 tosylate salt, BAY43-9006 tosylate salt, N-[4-Chloro-3-(trifluoromethyl)phenyl]-N′-[4-[2-(N-methylcarbamoyl)-4-pyridyloxy]phenyl]urea, 4-methylbenzenesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H16ClF3N4O3·C7H8SO3
CAS Number:
Molecular Weight:
637.03
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to very dark brown

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

FC(F)(F)c2c(ccc(c2)NC(=O)Nc3ccc(cc3)Oc4cc(ncc4)C(=O)NC)Cl.[S](=O)(=O)([O-])c1ccc(cc1)C.[H+]

InChI

1S/C21H16ClF3N4O3.C7H8O3S/c1-26-19(30)18-11-15(8-9-27-18)32-14-5-2-12(3-6-14)28-20(31)29-13-4-7-17(22)16(10-13)21(23,24)25;1-6-2-4-7(5-3-6)11(8,9)10/h2-11H,1H3,(H,26,30)(H2,28,29,31);2-5H,1H3,(H,8,9,10)

InChI key

IVDHYUQIDRJSTI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Orally active kinase inhibitor that exerts anticancer efficacy via targeting Raf-1 (c-Raf), b-Raf, VEGFR-2 (Flk-1, KDR), VEGFR-3, PDGFR-β, Flt-3, c-KIT, FGFR-1 (Flt-2) and RET.
Tosylate salt of the orally active kinase inhibitor Sorafenib (BAY 43-9006) that exerts broad-spectrum anticancer efficacy in vitro and in vivo via targeting b-Raf, c-Raf (Raf-1), as well as several receptor tyrosine kinases involved in neovascularization and tumor progression, including vascular endothelial growth factor receptors 2/3 (VEGFR-2/Flk-1/KDR, VEGFR-3), platelet-derived growth factor receptor-beta (PDGFR-β), Flt-3, c-KIT, FGFR-1 (Flt-2) and RET.

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Lact. - Repr. 1B - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Şükran Alpdemir et al.
IET nanobiotechnology, 14(7), 617-622 (2020-10-04)
This study aimed to develop sorafenib loaded magnetic microspheres for the treatment of hepatocellular carcinoma. To achieve this goal, superparamagnetic iron oxide nanoparticles (SPIONs) were synthesised and encapsulated in alginate microspheres together with an antineoplastic agent, sorafenib. In the study
Pan Liang et al.
International journal of pharmaceutics, 583, 119375-119375 (2020-04-29)
A novel nanocrystals delivery system of parthenolide (PTL) was designed to combined application with sorafenib (Sora) for advanced hepatocellular carcinoma (HCC) therapy, attempting to not only improve the poor aqueous solubility of PTL, but also enhance the synergistic therapeutic effects
Agnieszka Karbownik et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 130, 110530-110530 (2020-07-28)
Sorafenib (SR) is one of the most potent UGT (1A1, 1A9) inhibitors (in in vitro tests). The inhibition of UGT1A1 may cause hyperbilirubinaemia, whereas the inhibition of UGT1A9 and 1A1 may result in drug-drug interactions (DDIs). Tapentadol (TAP) is a
Agnieszka Karbownik et al.
Cancer chemotherapy and pharmacology, 85(6), 1039-1048 (2020-05-13)
Sorafenib is a multi-targeted tyrosine kinase inhibitor (TKI) used for the treatment of advanced renal cell carcinoma, hepatocellular carcinoma and radioactive iodine resistant thyroid carcinoma. Neoplastic diseases are the cause of pain, which may occur regardless of the stage of
Laurence Booth et al.
Biochemical pharmacology, 178, 114067-114067 (2020-06-07)
The combination of the multi-kinase and chaperone inhibitor sorafenib and the histone deacetylase inhibitor vorinostat in pancreatic cancer patients has proven to be a safe and efficacious modality (NCT02349867). We determined the evolutionary mechanisms by with pancreatic tumors become resistant

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service