Skip to Content
Merck
All Photos(1)

Key Documents

SML2442

Sigma-Aldrich

MS417

≥98% (HPLC)

Synonym(s):

(6S)-4-(4-Chlorophenyl)-2,3,9-trimethyl-6H-thieno[3,2-f][1,2,4]triazolo[4,3-a][1,4]diazepine-6-acetic acid methyl ester, GTPL 7512, Methyl 2-[(6S,Z)-4-(4-chlorophenyl)-2,3,9-trimethyl-6H-thieno[3,2-f][1,2,4]triazolo[4,3-a][1,4]diazepin-6-yl]acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H19ClN4O2S
CAS Number:
Molecular Weight:
414.91
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to very dark brown

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(OC)C[C@H]1C2=NN=C(C)N2C3=C(C(C)=C(C)S3)C(C4=CC=C(Cl)C=C4)=N1

InChI key

GGRCIHACOIMRKY-HNNXBMFYSA-N

Biochem/physiol Actions

MS417, a (+)-JQ1 analog, is a potent inhibitor of bromodomain-containing protein 4 (BRD4) that is highly specific for BRD4-BD1 and BRD4-BD2. MS417 binds with similar affinity to BRD4-BD1 and BRD4-BD2 (Kd =36.1, and 25.4 nM), while bulky tBu of JQ1 prevents tight binding to the cavity at BD1.
potent broad spectrum BET (Bromodomain and Extra-Terminal domain) bromodomain inhibitor

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service