Skip to Content
Merck
All Photos(1)

Key Documents

SML2278

Sigma-Aldrich

NDT9513727

≥98% (HPLC)

Synonym(s):

N,N-bis(1,3-Benzodioxol-5-ylmethyl)-1-butyl-2,4-diphenyl-1Himidazole-5-methanamine, NDT-9513727

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H35N3O4
CAS Number:
Molecular Weight:
573.68
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

CCCCN1C(CN(CC2=CC=C3OCOC3=C2)CC4=CC5=C(C=C4)OCO5)=C(N=C1C6=CC=CC=C6)C7=CC=CC=C7

InChI

1S/C36H35N3O4/c1-2-3-18-39-30(35(28-10-6-4-7-11-28)37-36(39)29-12-8-5-9-13-29)23-38(21-26-14-16-31-33(19-26)42-24-40-31)22-27-15-17-32-34(20-27)43-25-41-32/h4-17,19-20H,2-3,18,21-25H2,1H3

InChI key

ITACCRHKSPSKKL-UHFFFAOYSA-N

Biochem/physiol Actions

C5a receptor inverse agonist
NDT 9513727 is novel potent, selective and orally available C5a receptor inverse agonist.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robbin M Brodbeck et al.
The Journal of pharmacology and experimental therapeutics, 327(3), 898-909 (2008-08-30)
The complement system represents an innate immune mechanism of host defense that has three effector arms, the C3a receptor, the C5a receptor (C5aR), and the membrane attack complex. Because of its inflammatory and immune-enhancing properties, the biological activity of C5a
Yi-Min Huang et al.
Arthritis & rheumatology (Hoboken, N.J.), 67(10), 2780-2790 (2015-06-23)
Patients with antineutrophil cytoplasmic antibody-associated vasculitis (AAV) have a high frequency of venous thromboembolic events and a hypercoagulable state. As C5a-primed neutrophils play an important role in the development of AAV, we investigated whether C5a-induced neutrophil tissue factor (TF)-expressing microparticles
Nathan Robertson et al.
Nature, 553(7686), 111-114 (2018-01-05)
The complement system is a crucial component of the host response to infection and tissue damage. Activation of the complement cascade generates anaphylatoxins including C5a and C3a. C5a exerts a pro-inflammatory effect via the G-protein-coupled receptor C5a anaphylatoxin chemotactic receptor

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service