Skip to Content
Merck
All Photos(1)

Key Documents

SML1900

Sigma-Aldrich

Sobetirome

≥98% (HPLC)

Synonym(s):

2-[4-[[4-Hydroxy-3-(1-methylethyl)phenyl]methyl]-3,5-dimethylphenoxy]acetic acid, 3,5-Dimethyl-4-(4′-hydroxy-3′-isopropylbenzyl)phenoxyacetic acid, GC 1, GC-1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H24O4
CAS Number:
Molecular Weight:
328.40
UNSPSC Code:
51111800
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

−20°C

SMILES string

O(CC(=O)O)c1cc(c(c(c1)C)Cc2cc(c(cc2)O)C(C)C)C

InChI

1S/C20H24O4/c1-12(2)17-9-15(5-6-19(17)21)10-18-13(3)7-16(8-14(18)4)24-11-20(22)23/h5-9,12,21H,10-11H2,1-4H3,(H,22,23)

InChI key

QNAZTOHXCZPOSA-UHFFFAOYSA-N

Biochem/physiol Actions

Sobetirome is also termed as 2-(4-(4-(benzyloxy)-3-isopropylbenzyl)-3,5-dimethylphenoxy)acetic acid/ GC-1. It has an inner-ring and negatively charged carboxylate groups at physiological pH. Sobetirome is considered as a cholesterol lowering agent in humans.
Sobetirome is an orally active and potent agonist at thyroid hormone receptor (TR) β and TRα that displays selective tissue action.
Sobetirome is an orally active and potent agonist at thyroid hormone receptor (TR) β and TRα that displays selective tissue action. Sobetirome is devoid of thyrotoxic adverse effects on the heart, bone, and skeletal muscle.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jan Lammel Lindemann et al.
Expert opinion on therapeutic targets, 20(2), 145-149 (2015-11-14)
Sobetirome binds selectively to the main hepatic form of thyroid hormone (TH) receptor, TRβ1, compared to TRα1, which is principally responsible for thyrotoxic effects on heart, muscle and bone. Sobetirome also preferentially accumulates in liver. It was originally envisaged that
K Kannisto et al.
Atherosclerosis, 237(2), 544-554 (2014-12-03)
Thyroid hormone reduces plasma cholesterol and increases expression of low-density lipoprotein receptor (LDL-R) in liver, an effect mediated by thyroid receptor β (TRβ). The selective TRβ modulator GC-1 also enhances several steps in reverse cholesterol transport and can decrease serum
Sobetirome prodrug esters with enhanced blood?brain barrier permeability.
Placzek AT, et al.
Bioorganic & Medicinal Chemistry, 24(22), 5842-5854 (2016)
Ester-to-amide rearrangement of ethanolamine-derived prodrugs of sobetirome with increased blood-brain barrier penetration.
Ferrara SJ, et al.
Bioorganic & Medicinal Chemistry, 25(10), 2743-2753 (2017)
Andrew T Placzek et al.
Bioorganic & medicinal chemistry, 24(22), 5842-5854 (2016-10-26)
There is currently great interest in developing drugs that stimulate myelin repair for use in demyelinating diseases such as multiple sclerosis. Thyroid hormone plays a key role in stimulating myelination during development and also controls the expression of important genes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service