Skip to Content
Merck
All Photos(1)

Key Documents

SML1794

Sigma-Aldrich

Tenofovir disoproxil fumarate

≥98% (HPLC)

Synonym(s):

Bis(POC)PMPA fumarate, Bis{[(Isopropoxycarbonyl)oxy]methyl} ({[(2R)-1-(6-amino-9H-purin-9-yl)-2-propanyl]oxy}methyl)phosphonate fumarate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H30N5O10P · C4H4O4
CAS Number:
Molecular Weight:
635.51
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: 3 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

OC(/C=C/C(O)=O)=O.NC1=C2C(N(C[C@@H](C)OCP(OCOC(OC(C)C)=O)(OCOC(OC(C)C)=O)=O)C=N2)=NC=N1

InChI

1S/C19H30N5O10P.C4H4O4/c1-12(2)33-18(25)28-9-31-35(27,32-10-29-19(26)34-13(3)4)11-30-14(5)6-24-8-23-15-16(20)21-7-22-17(15)24;5-3(6)1-2-4(7)8/h7-8,12-14H,6,9-11H2,1-5H3,(H2,20,21,22);1-2H,(H,5,6)(H,7,8)/b;2-1+/t14-;/m1./s1

InChI key

VCMJCVGFSROFHV-WZGZYPNHSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Tenofovir disoproxil fumarate (tenofovir DF) is a nucleotide analogue with activity against retroviruses, such as human immunodeficiency virus (HIV)-1, HIV-2 and hepadnaviruses.
Tenofovir disoproxil fumarate is a prodrug of tenofovir, a nucleotide analogue reverse transcriptase inhibitor (nRTI) that causes premature termination of DNA transcription. In a T cell line and primary blood lymphocytes, the antiviral activity of tenofovir disoproxil was shown to be more than 100-fold greater than tenofovir because of its rapid intracellular uptake. Tenofovir disoproxil fumarate has been used alone and in various combinations for the prevention and treatment of HIV/AIDS and chronic hepatitis B infections and is on the World Health Organization′s List of Essential Medicines.
Tenofovir disoproxil fumarate is a prodrug of tenofovir, a nucleotide analogue reverse transcriptase inhibitor (nRTI); anti-retroviral.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - STOT RE 2

Target Organs

Bone,Kidney

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tenofovir Disoproxil Fumarate
Kearney B P, et al.
Clinical Pharmacokinetics, 43(9), 595-612 (2004)
B L Robbins et al.
Antimicrobial agents and chemotherapy, 42(3), 612-617 (1998-03-28)
Bis(isopropyloxymethylcarbonyl) 9-R-(2-phosphonomethoxypropyl)adenine [bis(POC)PMPA] has been identified as a novel prodrug of PMPA. The anti-human immunodeficiency virus activity of bis(POC)PMPA was >100-fold greater than that of PMPA in both an established T-cell line and primary peripheral blood lymphocytes. This improved efficacy
Tracy P Trang et al.
Expert opinion on drug safety, 15(9), 1287-1294 (2016-07-09)
Tenofovir disoproxil fumarate (TDF) and emtricitabine (FTC) are nucleoside reverse transcriptase inhibitors approved as pre-exposure prophylaxis (PrEP) against human immunodeficiency virus (HIV). Prophylactic TDF-based regimens have been shown to reduce the risk of HIV infection by 74 to 92% among
Kevin R Robertson et al.
AIDS (London, England), 30(15), 2315-2321 (2016-06-23)
The objective was to determine whether maraviroc (MVC) has unique neurocognitive benefits in the context of initial antiretroviral therapy (ART). Randomized, double-blind, placebo-controlled, 48-week trial. Participants were enrolled in US AIDS Clinical Trials Group clinical trial sites. Total 262 ART-naive
Elisa De Crignis et al.
eLife, 10 (2021-07-31)
The molecular events that drive hepatitis B virus (HBV)-mediated transformation and tumorigenesis have remained largely unclear, due to the absence of a relevant primary model system. Here we propose the use of human liver organoids as a platform for modeling

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service