Skip to Content
Merck
All Photos(1)

Key Documents

SML1463

Sigma-Aldrich

NHI-2

≥98% (HPLC)

Synonym(s):

1-Hydroxy-6-phenyl-4-(trifluoromethyl)-1H-Indole-2-carboxylic acid methyl ester, Methyl 1-hydroxy-6-phenyl-4-(trifluoromethyl)-1H-indole-2-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H12F3NO3
CAS Number:
Molecular Weight:
335.28
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

ON(C(C(OC)=O)=C1)C2=C1C(C(F)(F)F)=CC(C3=CC=CC=C3)=C2

InChI

1S/C17H12F3NO3/c1-24-16(22)15-9-12-13(17(18,19)20)7-11(8-14(12)21(15)23)10-5-3-2-4-6-10/h2-9,23H,1H3

InChI key

YPPFWRWCZNXINO-UHFFFAOYSA-N

Biochem/physiol Actions

NHI-2 is an inihbitor of Lactate dehydrogenase-A (LDH-A, LDHA), a key enzyme necessary to sustain glycolysis, the major pathway used by many cancer cells for cell growth and proliferation (the Warburg effect). NHI-2 has anti-glycolytic activity against a variety of cancer cells. NHI-2 inhibited cell growth of pancreatic cancer LPC006 and PANC-1 cells and enhanced the antiproliferative effects of Gemcitabine and caused apoptosis in colon cancer HCT116 cells.
NHI-2 is an inihbitor of Lactate dehydrogenase-A (LDH-A, LDHA).
NHI-2 possesses highr cell permeability.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carlotta Granchi et al.
Organic & biomolecular chemistry, 11(38), 6588-6596 (2013-08-30)
A head-to-head study of representative examples of N-hydroxyindole-2-carboxylates (NHI) and malonic derivatives (Mal) as LDH-A inhibitors was conducted, comparing the enzyme inhibition potency, cellular uptake, reduction of lactate production in cancer cells and anti-proliferative activity. Among the compounds tested, methyl
Maxime Alessandri et al.
iScience, 27(5), 109679-109679 (2024-04-24)
Epilepsy affects millions globally with a significant portion exhibiting pharmacoresistance. Abnormal neuronal activity elevates brain lactate levels, which prompted the exploration of its receptor, the hydroxycarboxylic acid receptor 1 (HCAR1) known to downmodulate neuronal activity in physiological conditions. This study

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service