Skip to Content
Merck
All Photos(1)

Key Documents

SML1224

Sigma-Aldrich

Torin2

≥98% (HPLC)

Synonym(s):

9-(6-Amino-3-pyridinyl)-1-[3-(trifluoromethyl)phenyl]-benzo[h]-1,6-naphthyridin-2(1H)-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H15F3N4O
CAS Number:
Molecular Weight:
432.40
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

FC(F)(F)c1cc(ccc1)[n]2c3c4c(ncc3cc[c]2=O)ccc(c4)c5cnc(cc5)N

InChI

1S/C24H15F3N4O/c25-24(26,27)17-2-1-3-18(11-17)31-22(32)9-6-16-13-29-20-7-4-14(10-19(20)23(16)31)15-5-8-21(28)30-12-15/h1-13H,(H2,28,30)

InChI key

GUXXEUUYCAYESJ-UHFFFAOYSA-N

Application

Torin2 has been used:
  • to examine its effect on LC3B-II levels in BORCS5- KO cells[1]
  • to determine its potent activity (IC50 less than 1 μM) against A2780-cis ovarian cancer cells[2]
  • in western blot analysis[3]

Biochem/physiol Actions

Torin2 is a highly potent and selective ATP-competitive mTOR inhibitor.
Torin2 is a highly potent and selective ATP-competitive mTOR inhibitor. Torin2 has an IC50 of 0.25 nM and 800-fold greater selectivity for mTOR than PI3K.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Small Molecules Identified from a Quantitative Drug Combinational Screen Resensitize Cisplatin's Response in Drug-Resistant Ovarian Cancer Cells
Sima N, et al.
Translational Oncology, 11(4), 1053-1064 (2018)
BORC coordinates encounter and fusion of lysosomes with autophagosomes
Jia R, et al.
Autophagy, 13(10), 1648-1663 (2017)
AMPK promotes induction of a tumor suppressor FLCN through activation of TFEB independently of mTOR
Collodet C, et al.
bioRxiv, 499921-499921 (2018)
Dan Song et al.
Journal of agricultural and food chemistry, 67(28), 7977-7985 (2019-04-02)
2-Amino-3,8-dimethylimidazo[4,5-f]quinoxaline (MeIQx), one of the most abundant heterocyclic aromatic amines (HAAs) found in the human diet, is primarily produced during high-temperature meat or fish cooking. While MeIQx has been investigated as a potential carcinogen, the cytotoxicity and related molecular mechanisms
Dan Song et al.
Environmental toxicology, 37(7), 1551-1562 (2022-03-04)
The aim of this study was to assess the protective effect and potential mechanism of melatonin against bisphenol A (BPA)-induced apoptosis and oxidative damage in FLK-BLV cells. The results showed that BPA reduced cell viability in a dose- and time-dependent

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service