Skip to Content
Merck
All Photos(1)

Key Documents

SML1124

Sigma-Aldrich

CPCCOEt

≥98% (HPLC)

Synonym(s):

7-Hydroxyiminocyclopropan[b]chromen-1α-carboxylic acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H13NO4
CAS Number:
Molecular Weight:
247.25
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(OCC)C(C1)(OC2=C/3C=CC=C2)C1C3=N/O

InChI

1S/C13H13NO4/c1-2-17-12(15)13-7-9(13)11(14-16)8-5-3-4-6-10(8)18-13/h3-6,9,16H,2,7H2,1H3/b14-11-

InChI key

FXCTZFMSAHZQTR-KAMYIIQDSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

CPCCOEt is a selective non-competitive metabotropic glutamate receptor mGluR1 antagonist with no agonist or antagonist activity at mGlu2, 4a, 5a, 7b, 8a or ionotropic receptors at concentrations of up to 100 μM.
CPCCOEt is a selective non-competitive metabotropic glutamate receptor mGluR1 antagonist.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bo Liu et al.
Brain research, 1704, 241-248 (2018-10-23)
Glioma is a primary brain tumor with high frequency and dismal prognosis. As there is no permanent cure available, identifying new therapy or mediator to augment the effectiveness of existing therapy is urgently needed. In the current study we tested
Carrie Bowman Dalley et al.
The Journal of pharmacology and experimental therapeutics, 367(1), 59-70 (2018-07-29)
Glioma refers to malignant central nervous system tumors that have histologic characteristics in common with glial cells. The most prevalent type, glioblastoma multiforme, is associated with a poor prognosis and few treatment options. On the basis of reports of aberrant
Valerie L Hedges et al.
Endocrinology, 159(3), 1328-1338 (2018-01-31)
Estrogens affect cerebellar activity and cerebellum-based behaviors. Within the adult rodent cerebellum, the best-characterized action of estradiol is to enhance glutamatergic signaling. However, the mechanisms by which estradiol promotes glutamatergic neurotransmission remain unknown. Within the mouse cerebellum, we found that
Xia Zhao et al.
Oxidative medicine and cellular longevity, 2020, 5173184-5173184 (2020-08-28)
Glutamate-induced neurotoxicity is involved in various neuronal diseases, such as Alzheimer's disease. We have previously reported that glutamate attenuated the survival signaling of insulin-like growth factor-1 (IGF-1) by N-methyl-D-aspartate receptors (NMDARs) in cultured cortical neurons, which is viewed as a
Christopher E Vaaga et al.
eLife, 9 (2020-03-25)
Innate defensive behaviors, such as freezing, are adaptive for avoiding predation. Freezing-related midbrain regions project to the cerebellum, which is known to regulate rapid sensorimotor integration, raising the question of cerebellar contributions to freezing. Here, we find that neurons of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service