Skip to Content
Merck
All Photos(1)

Key Documents

SML1055

Sigma-Aldrich

3-AP-Me

≥98% (HPLC)

Synonym(s):

2-[(3-Amino-2-pyridinyl)methylene]-N,N-dimethyl-hydrazinecarbothioamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H13N5S
CAS Number:
Molecular Weight:
223.30
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

CN(C)C(N/N=C/C1=C(N)C=CC=N1)=S

InChI

1S/C9H13N5S/c1-14(2)9(15)13-12-6-8-7(10)4-3-5-11-8/h3-6H,10H2,1-2H3,(H,13,15)/b12-6+

InChI key

DQVSEUUIXQTWRB-WUXMJOGZSA-N

Biochem/physiol Actions

3-AP-Me is a dimethyl derivative of the ribonucleotide reductase inhibitor 3-AP (SML0568). 3-AP-Me is a potent inducer of ER stress and the unfolded protein response in cancer cell lines, incuding IRE1, PERK and ATF6 activity. Treatment with 3-AP-Me induces CHOP expression, phosphorylation of eIF2α, up-regulation of ATF4 and Bim, and causes mitochondrial membrane depolarization and apoptosis.

Features and Benefits

This compound is a featured product for Apoptosis research. Click here to discover more featured Apoptosis products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service