Skip to Content
Merck
All Photos(1)

Documents

SML0897

Sigma-Aldrich

ALX 5407 hydrochloride

≥98% (HPLC)

Synonym(s):

ALX-5407 hydrochloride, N-[3-(4′-Fluorophenyl)-3-(4′-phenylphenoxy)propyl]sarcosine hydrochloride, NFPS hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H24NO3F·HCl
CAS Number:
Molecular Weight:
429.91
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

Cl.CN(CC[C@@H](Oc1ccc(cc1)-c2ccccc2)c3ccc(F)cc3)CC(O)=O

InChI

1S/C24H24FNO3.ClH/c1-26(17-24(27)28)16-15-23(20-7-11-21(25)12-8-20)29-22-13-9-19(10-14-22)18-5-3-2-4-6-18;/h2-14,23H,15-17H2,1H3,(H,27,28);1H/t23-;/m1./s1

InChI key

RPDGSZCYSJWQEE-GNAFDRTKSA-N

Looking for similar products? Visit Product Comparison Guide

Application

ALX 5407 hydrochloride has been used as a glycine transporter (GlyT1) inhibitor to test its effect on inhibitory postsynaptic current and N-methyl-D-aspartate receptor (NMDA)-mediated excitatory postsynaptic currents (EPSCs). It has also been used as a GlyT1 inhibitor to treat developmentally diminished NMDA receptor (Grin1D481N) mice to test its effect on the glycine site function.

Biochem/physiol Actions

ALX-5407 hydrochloride (NFPS hydrochloride) is a selective irreversible inhibitor of the glycine transporter GlyT1 with IC50 values of 3 nM for GlyT1 compared to 100 μM for GlyT2. ALX-5407 hydrochloride showed no activity at the inhibitory glycine receptor or glycine site of the NMDA receptor (IC50 > 100 mM).

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hyo-Jin Jeong et al.
British journal of pharmacology, 161(4), 925-935 (2010-09-24)
The arachidonyl-amino acid N-arachidonyl-glycine (NAGly) is an endogenous lipid, generated within the spinal cord and producing spinally mediated analgesia via non-cannabinoid mechanisms. In this study we examined the actions of NAGly on neurons within the superficial dorsal horn, a key
Viviane Labrie et al.
Psychopharmacology, 200(2), 217-230 (2008-07-04)
Schizophrenic patients demonstrate prominent negative and cognitive symptoms that are poorly responsive to antipsychotic treatment. Abnormal glutamatergic neurotransmission may contribute to these pathophysiological dimensions of schizophrenia. We examined the involvement of the glycine coagonist site on the N-methyl-D: -aspartate receptor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service