Skip to Content
Merck
All Photos(1)

Key Documents

SML0863

Sigma-Aldrich

A-1070722

≥98% (HPLC)

Synonym(s):

1-(7-Methoxyquinolin-4-yl)-3-[6-(trifluoromethyl)pyridin-2-yl]urea

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H13F3N4O2
CAS Number:
Molecular Weight:
362.31
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 15 mg/mL, clear

storage temp.

2-8°C

InChI

1S/C17H13F3N4O2/c1-26-10-5-6-11-12(7-8-21-13(11)9-10)22-16(25)24-15-4-2-3-14(23-15)17(18,19)20/h2-9H,1H3,(H2,21,22,23,24,25)

InChI key

VQPBIJGXSXEOCU-UHFFFAOYSA-N

Biochem/physiol Actions

A-1070722 is a potent brain-penetrant inhibitor of glycogen synthase kinase-3 (GSK-3) α and β isoforms with a Ki of 0.6 nM for both GSK-3α and GSK-3β. A-1070722 showed > 50-fold selectivity for GSK-3 over a panel of other kinases tested, including CDK family members. A-1070722 decreased phosphorylation of microtubule-associated protein Tau and protected rat primary cortical neurons against β amyloid and glutamate challenge.

Features and Benefits

This compound is featured on the GSK-3 and PKB/Akt pages of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service