Skip to Content
Merck
All Photos(1)

Key Documents

SML0824

Sigma-Aldrich

Lubeluzole dihydrochloride

≥98% (HPLC)

Synonym(s):

(aS)-4-(2-Benzothiazolylmethylamino)-a-[(3,4-difluorophenoxy)methyl]-1-piperidineethanol dihydrochloride, R 87926

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H25F2N3O2S · 2HCl
CAS Number:
Molecular Weight:
506.44
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

Fc1c(ccc(c1)OC[C@@H](O)CN2CCC(CC2)N(C)c3[s]c4c(n3)cccc4)F

InChI

1S/C22H25F2N3O2S/c1-26(22-25-20-4-2-3-5-21(20)30-22)15-8-10-27(11-9-15)13-16(28)14-29-17-6-7-18(23)19(24)12-17/h2-7,12,15-16,28H,8-11,13-14H2,1H3/t16-/m0/s1

InChI key

OZFSWVOEXHGDES-INIZCTEOSA-N

Biochem/physiol Actions

Lubeluzole is an NMDA antagonist.
Lubeluzole is an NMDA antagonist. Lubeluzole blocks glutamate release, as well as sodium and calcium gated ion channels. Lubeluzole displays neuroprotective properties in ischemia models.

Features and Benefits

This compound is featured on the Glutamate Receptors (Ion Channel Family) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jean-François Desaphy et al.
Experimental neurology, 255, 96-102 (2014-03-13)
Although the sodium channel blocker mexiletine is considered the first-line drug in myotonia, some patients experiment adverse effects, while others do not gain any benefit. Other antimyotonic drugs are thus needed to offer mexiletine alternatives. In the present study, we

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service