Skip to Content
Merck
All Photos(1)

Key Documents

SML0815

Sigma-Aldrich

YMU1

≥98% (HPLC)

Synonym(s):

4-[2-(4,6-Dimethyl-3-oxoisothiazolo[5,4-b]pyridin-2(3H)-yl)acetyl]-1-piperazinecarboxylic acid ethyl ester, Ethyl 4-(2-(4,6-dimethyl-3-oxoisothiazolo[5,4-b]pyridin-2(3H)-yl)acetyl)piperazine-1-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H22N4O4S
CAS Number:
Molecular Weight:
378.45
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to light brown

solubility

DMSO: 1 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

[s]1[n]([c](c3c1nc(cc3C)C)=O)CC(=O)N2CCN(CC2)C(=O)OCC

InChI

1S/C17H22N4O4S/c1-4-25-17(24)20-7-5-19(6-8-20)13(22)10-21-16(23)14-11(2)9-12(3)18-15(14)26-21/h9H,4-8,10H2,1-3H3

InChI key

XXUFNNVFZFVKGP-UHFFFAOYSA-N

Biochem/physiol Actions

YMU1 is a cell-permeable and nontoxic inhibitor of hTMPK (thymidylate kinase) that sensitizes tumor cells to doxorubicin in vitro and in vivo.
YMU1 is a cell-permeable and nontoxic inhibitor of hTMPK (thymidylate kinase).

Features and Benefits

This compound is a featured product for Apoptosis research. Click here to discover more featured Apoptosis products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service