Skip to Content
Merck
All Photos(1)

Key Documents

SML0647

Sigma-Aldrich

Phenazepam

≥97% (HPLC)

Synonym(s):

7-Bromo-5-(2-chlorophenyl)-1,3-dihydro-2H-1,4-benzodiazepin-2-one, 7-Bromo-5-(2-chlorophenyl)-1,3-dihydrobenzo[e]-1,4-diazepin-2-one, BD 98, Fenazepam

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10BrClN2O
CAS Number:
Molecular Weight:
349.61
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥97% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

−20°C

SMILES string

Brc1cc2c(cc1)NC(=O)CN=C2c3c(cccc3)Cl

InChI

1S/C15H10BrClN2O/c16-9-5-6-13-11(7-9)15(18-8-14(20)19-13)10-3-1-2-4-12(10)17/h1-7H,8H2,(H,19,20)

InChI key

CGMJQQJSWIRRRL-UHFFFAOYSA-N

Biochem/physiol Actions

Phenazepam is an agonist of the γ-Aminobutyric acid A (GABAA)-benzodiazepine receptor chloride channel complex.
Phenazepam is the benzodiazepine that exhibit anticonvulsive, anxiolytic, sedative and hypnotic effects in humans and experimental animals. Phenazepam is an agonist of the γ-Aminobutyric acid A (GABAA)-benzodiazepine receptor chloride channel complex.

Features and Benefits

This compound is featured on the GABAA Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Lact. - Muta. 1B - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

John Martin Corkery et al.
Human psychopharmacology, 27(3), 254-261 (2012-03-13)
Phenazepam (fenazepam; 7-bromo-5-(2-chlorophenyl)-1,3-dihydro-2H-1,4-benzodiazepin-2-one; PNZ, 'Bonsai') is a benzodiazepine developed in the former Soviet Union during the 1970s to treat neurological disorders, epilepsy, and alcohol withdrawal syndrome. Its recreational use appears to have increased over recent years. Because of the lack
Jon B Stephenson et al.
Journal of analytical toxicology, 37(1), 25-29 (2012-10-18)
Phenazepam use in the state of Georgia has increasingly become a trend for a drug market looking at new and different recreational drugs. This paper examines the psychomotor effects of phenazepam on individuals and their ability to operate a motor
[Evaluation of the effect of phenazepam and seduxen on the functional state of the cardiovascular system in the acute phase of an infarct].
G Lukhanana
Vrachebnoe delo, (10)(10), 15-18 (1983-10-01)
N Ia Golovenko et al.
Farmakologiia i toksikologiia, 43(2), 144-148 (1980-03-01)
The distribution pattern of 14C-phenazepam and some of its metabolites in the liver, brain and blood plasma at a prolonged (45 days) administration to rats does not differ from that after a single administration. The liver and brain demonstrated an
Peter D Maskell et al.
Journal of forensic and legal medicine, 19(3), 122-125 (2012-03-07)
In the past few years there has been concern in Western Europe and in the US about the rise in abuse of phenazepam, a benzodiazepine that was originally developed in the USSR in the mid- to late 1970s.(1-4) Although phenazepam

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service