Skip to Content
Merck
All Photos(1)

Key Documents

SML0602

Sigma-Aldrich

Propiverine hydrochloride

≥98% (HPLC)

Synonym(s):

α,α-Diphenyl-α-propoxyacetic acid hydrochloride 1-methyl-4-piperidyl ester, α-Phenyl-α-propoxybenzeneacetic acid 1-methyl-4-piperidinyl ester hydrochloride, (1-Methylpiperidin-1-ium-4-yl) 2,2-diphenyl-2-propoxyacetate chloride, 1-Methyl-4-piperidyl diphenylpropoxyacetate hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H29NO3 · HCl
CAS Number:
Molecular Weight:
403.94
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

[Cl-].[N+H]1(CCC(CC1)OC(=O)C(OCCC)(c3ccccc3)c2ccccc2)C

InChI

1S/C23H29NO3.ClH/c1-3-18-26-23(19-10-6-4-7-11-19,20-12-8-5-9-13-20)22(25)27-21-14-16-24(2)17-15-21;/h4-13,21H,3,14-18H2,1-2H3;1H

InChI key

KFUJMHHNLGCTIJ-UHFFFAOYSA-N

General description

Propiverine hydrochloride drug is a tertiary amine with spasmolytic and analgesic properties. This drug exerts neurotropic and musculotropic effects on the urinary bladder and smooth muscles. Propiverine hydrochloride has a half-life of 20 hours.

Biochem/physiol Actions

Propiverine is an antimuscarinic drug used for the treatment of overactive blaqdder and urinary incontinence. Propiverine and/or its metabolites also have calcium channel blocking properties in addition to antimuscarinic activity, which may contribute to the clinical activity.
Propiverine is an antimuscarinic drug used for the treatment of overactive blaqdder and urinary incontinence.

Features and Benefits

This compound is featured on the Acetylcholine Receptors (Muscarinic) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Werner Siegmund et al.
Journal of clinical pharmacology, 52(5), 681-690 (2011-05-10)
The muscarinic receptor antagonist propiverine is unique insofar as extended-release (ER) tablets are of higher bioavailability than immediate-release (IR) tablets; this is caused by lower "first-pass" elimination of propiverine via CYP3A4 and efflux transporters in the distal small intestine and
Yasue Kubota et al.
Neurourology and urodynamics, 30(7), 1309-1314 (2011-05-12)
To evaluate the relationship between treatment-related changes in Overactive Bladder Symptom Scores (OABSS) and health-related quality of life (HRQOL) questionnaires. Ninety-five patients with OAB symptoms were enrolled. All patients completed the OABSS, International Prostate Symptom Score (IPSS)-Quality Of Life (QOL)
Editorial comment from Dr Dmochowski to propiverine hydrochloride in Japanese patients with overactive bladder: a randomized, double-blind, placebo-controlled trial.
Roger R Dmochowski
International journal of urology : official journal of the Japanese Urological Association, 18(5), 374-374 (2011-03-02)
Daniela Schultz-Lampel et al.
Deutsches Arzteblatt international, 108(37), 613-620 (2011-10-07)
Urinary incontinence (bedwetting, enuresis) is the commonest urinary symptom in children and adolescents and can lead to major distress for the affected children and their parents. Physiological and non-physiological types of urinary incontinence are sometimes hard to tell apart in
Tetsuo Ogata et al.
Chemical & pharmaceutical bulletin, 60(8), 976-984 (2012-08-07)
The aim of the present study was to mask the bitterness of propiverine hydrochloride (P-4) by converting it to propiverine free base. Fine granules comprising the free base, which was converted from P-4 by desalination, were prepared. By using Fourier

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service