Skip to Content
Merck
All Photos(1)

Documents

SML0593

Sigma-Aldrich

CGP52432

≥98% (HPLC)

Synonym(s):

[3-[[(3,4-Dichlorophenyl)methyl]amino]propyl](diethoxymethyl)-phosphinic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H24Cl2NO4P
CAS Number:
Molecular Weight:
384.24
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 1 mg/mL, clear (warmed)

storage temp.

2-8°C

InChI

1S/C15H24Cl2NO4P/c1-3-21-15(22-4-2)23(19,20)9-5-8-18-11-12-6-7-13(16)14(17)10-12/h6-7,10,15,18H,3-5,8-9,11H2,1-2H3,(H,19,20)

InChI key

GJZVQXWEIYRHBE-UHFFFAOYSA-N

Application

CGP52432 has been used as a γ-aminobutyric acid B (GABAB) antagonist:
  • to study its effects on the simulation of the onset of status epilepticus (SE) in mice
  • for voltage-clamp recording in mice neurons
  • to study its effects on the GABAB receptor-mediated neurotransmission in guinea pig hippocampus

Biochem/physiol Actions

CGP52432 is a very potent antagonist of GABAB receptors. (IC50 = 85 nM).
CGP52432 participates in inhibiting the glycine overflow in the hippocampus.

Features and Benefits

This compound is featured on the GABAB Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Other Notes

Do not freeze.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A E Matukhno et al.
Zhurnal vysshei nervnoi deiatelnosti imeni I P Pavlova, 62(3), 372-382 (2012-08-16)
By means of local microapplication of GABA, picrotoxin and CGP 52432, different roles of GABAA and GABAB receptors in the geneses of primary and secondary components of evoked potentials in the somatosensory barrel cortex of rats were shown. The authors
Chunguang Zhang et al.
Endocrinology, 150(5), 2388-2394 (2009-01-24)
Gamma-aminobutyric acid (GABA) is one of the most important neurotransmitters that regulate the excitability of GnRH neurons. Numerous studies have shown that GABA activates Cl(-) currents in GnRH neurons, and these effects are antagonized by GABA(A) receptor antagonists. The GABA(B)
Kazuo Kitamura et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 31(30), 10847-10858 (2011-07-29)
Cerebellar Purkinje cells have one of the most elaborate dendritic trees in the mammalian CNS, receiving excitatory synaptic input from a single climbing fiber (CF) and from ∼200,000 parallel fibers. The dendritic Ca(2+) signals triggered by activation of these inputs
Shuhei Kobuchi et al.
European journal of pharmacology, 623(1-3), 113-118 (2009-09-22)
Enhanced renal sympathetic nerve activity during ischemic period and the renal venous norepinephrine overflow after reperfusion play important roles in the development of ischemic acute kidney injury. We investigated the effect of gamma-aminobutyric acid (GABA), an inhibitory neurotransmitter mainly in
Cristina Romei et al.
Pharmacological research, 61(6), 547-552 (2010-02-09)
GABA(B) receptors mediate inhibition of neurotransmitter exocytosis from nerve endings. Unexpectedly, the well known GABA(B) receptor antagonist CGP35348 and, in part, the compound CGP52432, are now found to inhibit on their own the K(+)-evoked exocytosis of glycine when added at

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service