Skip to Content
Merck
All Photos(1)

Key Documents

SML0437

Sigma-Aldrich

Oxandrolone

≥98% (HPLC)

Synonym(s):

17β-Hydroxy-17α-methyl-2-oxa-5α-androstan-3-one, 8075CB, NSC 67068, Oxandren, Provitar, SC 11585, Vasorome

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H30O3
CAS Number:
Molecular Weight:
306.44
EC Number:
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D -19 to -26°, c = 1 (CDCl3)

drug control

USDEA Schedule IIIN; regulated under CDSA - not available from Sigma-Aldrich Canada

color

white to beige

solubility

DMSO: 2 mg/mL (clear solution; warmed)

storage temp.

2-8°C

SMILES string

C[C@]1(O)CC[C@H]2[C@@H]3CC[C@H]4CC(=O)OC[C@]4(C)[C@H]3CC[C@]12C

InChI

1S/C19H30O3/c1-17-11-22-16(20)10-12(17)4-5-13-14(17)6-8-18(2)15(13)7-9-19(18,3)21/h12-15,21H,4-11H2,1-3H3/t12-,13+,14-,15-,17-,18-,19-/m0/s1

InChI key

QSLJIVKCVHQPLV-PEMPUTJUSA-N

Gene Information

human ... AR(367)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Oxandrolone is a synthetic anabolic steroid. It is is a non-aromatizable androgen with no estrogenic effects and with mild androgenic activity. Clinical uses of oxandrolone include to promote weight gain after weight loss following extensive surgery or chronic infections or trauma, to offset the protein catabolism associated with prolonged administration of corticosteroids, to relieve bone pain frequently accompanying osteoporosis, and to treat Turner′s syndrome in girls.
Oxandrolone is an anabolic steroid.

Features and Benefits

This compound is featured on the Nuclear Receptors (Steroids) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rhonda Orr et al.
Drugs, 64(7), 725-750 (2004-03-18)
There has been increasing interest in the development of effective agents that can be safely used to promote anabolism in the clinical setting for patients with chronic wasting conditions as well as in the prevention and treatment of frailty associated
Martha P D Zeger et al.
Hormone research in paediatrics, 75(1), 38-46 (2010-08-25)
Untreated girls with Turner syndrome (TS) have growth failure, and adult height is, on average, 20 cm less than predicted height. Treatment with growth hormone (GH) is now standard of care. The objective of this study was to investigate the
Yesenia Rojas et al.
Expert opinion on pharmacotherapy, 13(17), 2485-2494 (2012-11-06)
The worldwide occurrence of burn injuries remains high despite efforts to reduce injury incidence through public awareness campaigns and improvements in living conditions. In 2004, almost 11 million people experienced burns severe enough to warrant medical treatment. Advances over the
Leonie A Menke et al.
Journal of voice : official journal of the Voice Foundation, 25(5), 602-610 (2010-10-26)
Oxandrolone (Ox) increases height gain but may also cause voice deepening in growth hormone (GH)-treated girls with Turner syndrome (TS). We assessed the effect of Ox on objective and subjective speaking voice frequency in GH-treated girls with TS. A multicenter
Leonie A Menke et al.
Hormone research in paediatrics, 75(2), 115-122 (2010-09-04)
The weak androgen oxandrolone (Ox) may increase height but may also affect glucose metabolism in girls with Turner syndrome (TS). In a randomized, placebo-controlled, double-blind study, we assessed the effect of Ox at a dosage of either 0.06 or 0.03

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service