Skip to Content
Merck
All Photos(1)

Key Documents

SML0314

Sigma-Aldrich

CGP77675

≥98% (HPLC)

Synonym(s):

1-(2-{4-[4-Amino-5-(3-methoxyphenyl)-7H-pyrrolo[2,3-d]pyrimidin-7-yl]phenyl}ethyl)piperidin-4-ol, 1-(4-(4-Amino-5-(3-methoxyphenyl)-7H-pyrrolo[2,3-d]pyrimidin-7-yl)phenethyl)piperidin-4-ol, CGP 77675, CGP-77675

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H29N5O2
CAS Number:
Molecular Weight:
443.54
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: >10 mg/mL

storage temp.

2-8°C

SMILES string

COc1cccc(c1)-c2cn(-c3ccc(CCN4CCC(O)CC4)cc3)c5ncnc(N)c25

InChI

1S/C26H29N5O2/c1-33-22-4-2-3-19(15-22)23-16-31(26-24(23)25(27)28-17-29-26)20-7-5-18(6-8-20)9-12-30-13-10-21(32)11-14-30/h2-8,15-17,21,32H,9-14H2,1H3,(H2,27,28,29)

InChI key

WUPXZZWTHIZICK-UHFFFAOYSA-N

General description

CGP77675 belongs to the pyrrolopyrimidine class of inhibitors.

Application

CGP77675 has been used in the generation of mouse expanded potential stem cells.

Biochem/physiol Actions

CGP77675 is a potent Src kinase inhibitor. CGP77675 is selective toward other protein kinases including Cdc2 (>500-fold), epidermal growth factor (EGF) receptor (7.5-fold), vascular endothelial growth factor receptor (>50-fold), and for v-Abl (15-fold) and focal adhesion kinase (Fak) (>25-fold).
CGP77675 prevents hypercalcemia stimulated by interleukin 1β. It is known to offer protection against bone loss in ovariectomized mice. CGP77675 is known for its use in cancer treatment.

Features and Benefits

This compound is a featured product for Kinase Phosphatase Biology research. Click here to discover more featured Kinase Phosphatase Biology products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Osteoporosis, 798-798 (2001)
Targeted Interference with Signal Transduction Events, 177-177 (2007)
Ingun Heiene Tveteraas et al.
Journal of experimental & clinical cancer research : CR, 31, 72-72 (2012-09-13)
It is important to understand the mechanisms by which the cells integrate signals from different receptors. Several lines of evidence implicate epidermal growth factor (EGF) receptor (EGFR) in the pathophysiology of hepatocarcinomas. Data also suggest a role of prostaglandins in
Wataru Hirata et al.
Experimental animals (2021-09-22)
In mammals, sexual fate is determined by the chromosomes of the male and female gametes during fertilization. Males (XY) or females (XX) are produced when a sperm containing a Y or X-chromosome respectively fertilizes an X-chromosome-containing unfertilized egg. However, sexing
Establishment of mouse expanded potential stem cells
Yang J, et al.
Nature, 550(7676), 393-393 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service