Skip to Content
Merck
All Photos(1)

Key Documents

SML0269

Sigma-Aldrich

Nitisinone

≥95% (HPLC)

Synonym(s):

2-(2-Nitro-4-trifluoromethylbenzoyl)-1,3-cyclohexanedione, 2-[2-Nitro-4-(trifluoromethyl)benzoyl]cyclohexane-1,3-dione, NTBC, Nitisone, SC 0735

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H10F3NO5
CAS Number:
Molecular Weight:
329.23
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

powder

color

white to brown

solubility

DMSO: ≥5 mg/mL

storage temp.

−20°C

SMILES string

[O-][N+](=O)c1cc(ccc1C(=O)C2C(=O)CCCC2=O)C(F)(F)F

InChI

1S/C14H10F3NO5/c15-14(16,17)7-4-5-8(9(6-7)18(22)23)13(21)12-10(19)2-1-3-11(12)20/h4-6,12H,1-3H2

InChI key

OUBCNLGXQFSTLU-UHFFFAOYSA-N

Gene Information

human ... HPD(3242)

Looking for similar products? Visit Product Comparison Guide

Application

Nitisinone has been used:
  • to induce liver injury
  • to treat Ixodes scapularis tick cells to inhibit the activity of hydroxyphenylpyruvate dioxygenase
  • in supplemented water to block the accumulation of toxic metabolites in human hepatocyte engrafted mice
  • to study its effect on bacterial pyomelanin production

Biochem/physiol Actions

Nitisinone is a competitive inhibitor that reversibly inhibits 4-Hydroxyphenylpyruvate oxidase (dioxygenase). Nitisinone is used in the treatment of hereditary tyrosinemia type 1, where it blocks the degradation of tyrosine into harmful substances.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jean Larochelle et al.
Molecular genetics and metabolism, 107(1-2), 49-54 (2012-08-14)
Hepatorenal tyrosinemia (HT1, fumarylacetoacetate hydrolase deficiency, MIM 276700) can cause severe hepatic, renal and peripheral nerve damage. In Québec, HT1 is frequent and neonatal HT1 screening is practiced. Nitisinone (NTBC, Orfadin ®) inhibits tyrosine degradation prior to the formation of
Andrew J Preston et al.
Annals of the rheumatic diseases, 73(1), 284-289 (2013-03-21)
Alkaptonuria (AKU) is a rare metabolic disease caused by deficiency of homogentisate 1,2 dioxygenase, an enzyme involved in tyrosine catabolism, resulting in increased circulating homogentisic acid (HGA). Over time HGA is progressively deposited as a polymer (termed ochronotic pigment) in
Pieter-Paul Schauwvlieghe et al.
Cornea, 32(1), 91-94 (2012-04-13)
To describe the confocal microscopic findings in a patient with hereditary tyrosinemia type I (HT-I) treated with 2-(2-nitro-4-trifluoromethylbenzoyl)-1,3-cyclohexanedione (NTBC) who developed corneal crystals. In this case study, we describe the confocal microscopic findings in a boy, who was diagnosed with
Eva Thimm et al.
Molecular genetics and metabolism, 102(2), 122-125 (2010-11-30)
Psychomotor impairment has been described in hypertyrosinemia types II and III (HT III). Only recently cognitive deficits have also been reported in hypertyrosinemia type I (HT I). The pathogenic mechanisms responsible are unknown. Since implementation of 2-(2-nitro-4-trifluoromethylbenzoyl)-1,3-cyclohexanedione (NTBC, Nitisinone (Swedish
Saikat Santra et al.
Expert opinion on pharmacotherapy, 9(7), 1229-1236 (2008-04-22)
Hereditary tyrosinaemia type 1 is a rare inherited metabolic condition, which leads to a fatal multisystemic disease in childhood. Since 1992, nitisinone - a compound developed from work on triketone herbicides - has become an effective pharmacological treatment by inhibiting

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service