Skip to Content
Merck
All Photos(1)

Key Documents

SML0216

Sigma-Aldrich

Brinzolamide

≥98% (HPLC)

Synonym(s):

(5R)-5-Ethylamino-3-(3-methoxypropyl)-2,2-dioxo-2?6,9-dithia-3-azabicyclo[4.3.0]nona-7,10-diene-8-sulfonamide, (R)- 4-(ethylamino)-3,4-dihydro-2-(3-methoxypropyl)-2H-Thieno[3,2-e]-1,2-thiazine-6-sulfonamide 1,1-dioxide, AL 4862

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H21N3O5S3
CAS Number:
Molecular Weight:
383.51
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: ≥10 mg/mL

storage temp.

−20°C

SMILES string

CCN[C@H]1CN(CCCOC)S(=O)(=O)c2sc(cc12)S(N)(=O)=O

InChI

1S/C12H21N3O5S3/c1-3-14-10-8-15(5-4-6-20-2)23(18,19)12-9(10)7-11(21-12)22(13,16)17/h7,10,14H,3-6,8H2,1-2H3,(H2,13,16,17)/t10-/m0/s1

InChI key

HCRKCZRJWPKOAR-JTQLQIEISA-N

Gene Information

human ... CA2(760)

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Brinzolamide is a small molecular weight compound that has an ability to bind melanin. This drug is used in ocular therapy.

Application

Brinzolamide has been used as a melanin binding compound or drug in melanin binding assays. It has also been used as a carbonic anhydrase inhibitor (CAI).

Biochem/physiol Actions

Brinzolamide is a carbonic anhydrase II inhibitor
Brinzolamide is a carbonic anhydrase II inhibitor used to lower intraocular pressure.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gemma Caterina Maria Rossi et al.
Expert opinion on pharmacotherapy, 12(16), 2425-2431 (2011-06-18)
To assess ocular surface status and tolerability after switching glaucoma patients from dorzolamide/timolol to brinzolamide/timolol fixed combination (FC). Six-month, multicenter, open-label, prospective study that switched 72 patients from dorzolamide/timolol to brinzolamide/timolol FC. Intraocular pressure (IOP), tear film break-up-time (TF-BUT), fluorescein
Ivan Goldberg et al.
Journal of glaucoma, 21(1), 55-59 (2010-11-05)
To compare the efficacy of brinzolamide versus placebo when added to travoprost/timolol fixed combination (TTFC) in uncontrolled patients. This was a prospective, double-masked, randomized, placebo-controlled, parallel comparison of ocular hypertensive or primary open-angle glaucoma patients. Patients treated with a prostaglandin-based
To Be or Not to Be Protonated: Atomic Details of Human Carbonic Anhydrase-Clinical Drug Complexes by Neutron Crystallography and Simulation.
Kovalevsky A, et al.
Structure, 26(3), 383-390 (2018)
Gemma Caterina Maria Rossi et al.
Expert opinion on pharmacotherapy, 12(5), 685-690 (2011-03-10)
To examine the impact of switching glaucoma patients from timolol 0.5% to brinzolamide 1%/timolol 0.5% fixed combination (Brinz/Tim FC) on quality of life and on ocular surface status; to assess efficacy after the switch. 6-month, multicenter, open-label, prospective, switch study
John H K Liu et al.
Ophthalmology, 118(10), 1995-2000 (2011-06-17)
A core assumption for the 1-eye therapeutic trial of ocular hypotensive medications is the symmetrical reduction of intraocular pressure (IOP) in paired eyes. This assumption was evaluated for 24-hour IOP reduction in patients who underwent monotherapy or adjunctive therapy. Database

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service