Skip to Content
Merck
All Photos(1)

Key Documents

SML0059

Sigma-Aldrich

BLT-1

≥98% (HPLC)

Synonym(s):

2-Hexyl-1-cyclopentanone thiosemicarbazone, 33M20, BLT1, Block lipid transport-1, MIT 9952-53

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H23N3S
CAS Number:
Molecular Weight:
241.40
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to tan

solubility

DMSO: ≥13 mg/mL

storage temp.

2-8°C

SMILES string

CCCCCCC1CCC\C1=N/NC(N)=S

InChI

1S/C12H23N3S/c1-2-3-4-5-7-10-8-6-9-11(10)14-15-12(13)16/h10H,2-9H2,1H3,(H3,13,15,16)/b14-11+

InChI key

OWGUSBISUVLUJF-SDNWHVSQSA-N

Application

BLT-1 (Block lipid transport-1) has been used as an inhibitor of the SR-BI (scavenger receptor, class B, type I) mediated lipid transfer to determine if miR-223-3p export from polymorphonuclear neutrophils (PMNs) to high-density lipoproteins (HDLs) is dependent on SR-BI-mediated lipid flux. It has also been used as a SR-BI inhibitor to study the effect of HDL on osteocalcin (OCN) expression in monocyte cell lines through SR-B1.
BLT-1 may be used to study cell signaling pathways that have role in cholesterol influx and efflux.

Biochem/physiol Actions

Block lipid transport-1 (BLT-1) is a specific inhibitor of the SR-BI (Scavenger receptor, class B, type I) mediated lipid transfer. The compound inhibits both cellular selective lipid uptake of HDL cholesteryl ether and efflux of cellular cholesterol to HDL.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fumiyuki Sasaki et al.
PloS one, 12(9), e0185133-e0185133 (2017-09-19)
Leukotriene B4 (LTB4) receptor 1 (BLT1) is a G protein-coupled receptor expressed in various leukocyte subsets; however, the precise expression of mouse BLT1 (mBLT1) has not been reported because a mBLT1 monoclonal antibody (mAb) has not been available. In this
Yue-Hua Feng et al.
Lipids in health and disease, 17(1), 200-200 (2018-08-27)
Scavenger receptor BI (SR-BI) is a classic high-density lipoprotein (HDL) receptor, which mediates selective lipid uptake from HDL cholesterol esters (HDL-C). Apolipoprotein M (ApoM), as a component of HDL particles, could influence preβ-HDL formation and cholesterol efflux. The aim of
Ceren Emre et al.
Brain pathology (Zurich, Switzerland), 30(3), 614-640 (2020-01-09)
Neuroinflammation is a key element of AD pathology and conceivably a result of a disturbed resolution. Resolution of inflammation is an active process which is strictly orchestrated following the acute inflammatory response after removal of the inflammatory stimuli. Acute inflammation
High density lipoprotein modulates osteocalcin expression in circulating monocytes: a potential protective mechanism for cardiovascular disease in type 1 diabetes
Maddaloni E, et al.
Cardiovascular Diabetology, 16(1), 1-11 (2017)
Ivana Halova et al.
Frontiers in immunology, 3, 119-119 (2012-06-02)
Migration of mast cells is essential for their recruitment within target tissues where they play an important role in innate and adaptive immune responses. These processes rely on the ability of mast cells to recognize appropriate chemotactic stimuli and react

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service