Skip to Content
Merck
All Photos(1)

Key Documents

SMB00509

Sigma-Aldrich

1,2-Dimyristoyl-sn-glycerol

≥95%

Synonym(s):

(S)-1,2-Dimyristin, (S)-Glycerol 1,2-dimyristate, 1,2-Ditetradecanoyl-sn-glycerol, DMG

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H60O5
CAS Number:
Molecular Weight:
512.81
Beilstein:
1808871
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic (organic)

Quality Level

Assay

≥95%

form

powder

functional group

ketone

lipid type

neutral glycerides

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCCCC(OC[C@H](CO)OC(CCCCCCCCCCCCC)=O)=O

InChI

1S/C31H60O5/c1-3-5-7-9-11-13-15-17-19-21-23-25-30(33)35-28-29(27-32)36-31(34)26-24-22-20-18-16-14-12-10-8-6-4-2/h29,32H,3-28H2,1-2H3/t29-/m0/s1

InChI key

JFBCSFJKETUREV-LJAQVGFWSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

1,2-Dimyristoyl-sn-glycerol (1,2-ditetradecanoyl-sn-glycerol) may be used as a reactant in the preparation of 1,2-[13,13-2H2]dimyristoyl-sn-glycero-3-phosphoethanolamine and sodium 1,2-ditetradecanoyl-sn-glycer-3-yl phenyl phosphate.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dynamic structure and phase behavior of dimyristoylphosphatidylethanolamine bilayers studied by deuterium nuclear magnetic resonance.
Marsh D, et al.
Biochemistry, 22(12), 3023-3026 (1983)
Orientation of the headgroup of phosphatidylinositol in a model biomembrane as determined by neutron diffraction.
Bradshaw JP, et al.
Biochemistry, 38(26), 8393-8401 (1999)
L D'Abundo et al.
Oncogene, 36(47), 6617-6626 (2017-08-08)
Dysregulation of microRNAs (miRNAs) plays an important role in the pathogenesis of chronic lymphocytic leukemia (CLL). The Eμ-TCL1 transgenic mouse develops a form of leukemia that is similar to the aggressive type of human B-CLL, and this valuable model has
Suvasini Ramaswamy et al.
Proceedings of the National Academy of Sciences of the United States of America, 114(10), E1941-E1950 (2017-02-17)
Safe and efficient delivery of messenger RNAs for protein replacement therapies offers great promise but remains challenging. In this report, we demonstrate systemic, in vivo, nonviral mRNA delivery through lipid nanoparticles (LNPs) to treat a Factor IX (
Kaoru Nomura et al.
Biophysical journal, 117(1), 99-110 (2019-06-06)
After a nascent chain of a membrane protein emerges from the ribosomal tunnel, the protein is integrated into the cell membrane. This process is controlled by a series of proteinaceous molecular devices, such as signal recognition particles and Sec translocons.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service