Skip to Content
Merck
All Photos(1)

Key Documents

SMB00295

Sigma-Aldrich

Ruscogenin

≥98% (HPLC)

Synonym(s):

(1β,3β,25R)-Spirost-5-ene-1,3-diol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H42O4
CAS Number:
Molecular Weight:
430.62
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥98% (HPLC)

form

powder

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

C[C@H](CC1)CO[C@]21O[C@@]3([H])C[C@@]4([H])[C@]5([H])CC=C6C[C@@H](O)C[C@@H](O)[C@]6(C)[C@@]5([H])CC[C@]4(C)[C@@]3([H])[C@@H]2C

InChI

1S/C27H42O4/c1-15-7-10-27(30-14-15)16(2)24-22(31-27)13-21-19-6-5-17-11-18(28)12-23(29)26(17,4)20(19)8-9-25(21,24)3/h5,15-16,18-24,28-29H,6-14H2,1-4H3/t15-,16+,18-,19-,20+,21+,22+,23-,24+,25+,26+,27-/m1/s1

InChI key

QMQIQBOGXYYATH-IDABPMKMSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ruscogenin is a major steroidal saponin found in the root of Ophiopogon japonicus. It was first isolated from Ruscus aculeatus.

Application

Ruscogenin has been used to study its therapeutic effects on Crohn′s disease (CD)-like colitis both in vitro and in vivo studies.

Biochem/physiol Actions

Ruscogenin is a well-known compound for down-regulating nuclear factor kappa B (NF-κB), intercellular adhesion molecule-1 (ICAM-1), tissue factor (TF), inducible NO synthase (iNOS), and modulating inflammatory pathways. It displays anti-inflammatory properties by inhibiting lipopolysaccharide (LPS)-induced acute lung injury (ALI), and improving diabetic nephropathy in in vivo studies. Ruscogenin also exerts neuroprotective properties and protects the brain against ischemic injury in mouse model. It elicits anti-tumor properties by inducing ferroptosis in pancreatic cancer cells in both in vitro and in vivo studies.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ya-Lin Huang et al.
Journal of pharmacological sciences, 108(2), 198-205 (2008-10-24)
Ruscogenin (RUS), first isolated from Ruscus aculeatus, also a major steroidal sapogenin of traditional Chinese herb Radix Ophiopogon japonicus, has been found to exert significant anti-inflammatory and anti-thrombotic activities. Our previous studies suggested that ruscogenin remarkably inhibited adhesion of leukocytes
C J O'Brien et al.
World journal of surgery, 16(2), 222-226 (1992-03-01)
This retrospective study examines the experience of the Sydney Melanoma Unit in the management of cervical lymph nodes among patients with cutaneous melanoma of the head and neck. From 1960 to 1990, 397 patients had neck dissections for cutaneous malignant
A J Cochran et al.
World journal of surgery, 16(2), 214-221 (1992-03-01)
One of the most difficult decisions in the management of patients with melanoma remains whether or not to dissect electively the regional lymph nodes of patients with high risk primary tumors. There is abundant evidence that a proportion of such
J Vermeiren et al.
Acta orthopaedica Belgica, 58(1), 88-90 (1992-01-01)
Partial and sometimes full-thickness rotator cuff tears can be managed by arthroscopic debridement and anterior acromioplasty. The recent literature on shoulder arthroscopy does not however mention any alternative operation techniques for this very common lesion. We present a case of
Domains of muscarinic acetylcholine receptors that confer specificity of G protein coupling.
T I Bonner
Trends in pharmacological sciences, 13(2), 48-50 (1992-02-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service