Skip to Content
Merck
All Photos(1)

Key Documents

SMB00103

Sigma-Aldrich

Abietin

≥90% (LC/MS-ELSD)

Synonym(s):

Coniferin, Coniferoside, Coniferyl alcohol β-D-glucoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H22O8
CAS Number:
Molecular Weight:
342.34
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

Assay

≥90% (LC/MS-ELSD)

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

COc1cc(C\C=C\O)ccc1O[C@@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O

InChI

1S/C16H22O8/c1-22-11-7-9(3-2-6-17)4-5-10(11)23-16-15(21)14(20)13(19)12(8-18)24-16/h2,4-7,12-21H,3,8H2,1H3/b6-2+/t12-,13-,14+,15-,16-/m1/s1

InChI key

XQIJIPVRXMWYLN-GLIXSRQJSA-N

General description

Abietin, also known as Coniferin, is a glucoside of coniferyl alcohol that is also found in the water root extract of Angelica archangelica subsp. litoralis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mei-hua Han et al.
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica, 32(17), 1755-1759 (2007-11-13)
To study the bioactive constituents of the fresh rhizome of Pinellia ternata, and provide the scientific basis for quality control. Various chromatographic techniques were used to separate and purify the chemical constituents, and their chemical structures were determined on the
K Ito et al.
Bioscience, biotechnology, and biochemistry, 64(3), 584-587 (2000-05-10)
The fraction containing glycosidic components was separated from flue-cured tobacco (Nicotiana tabacum L.) leaves by a facile method. Some components of the fraction were isolated and elucidated to be syringin, coniferin, cichoriin, benzyl-beta-D-glucoside, Blumenol A-beta-D-glucoside, and 5,6-epoxy-5,6-dihydro-3-hydroxy-beta-ionyl-beta-D-glucoside. Syringin and coniferin
Han-wei Lin et al.
Biotechnology letters, 25(7), 521-525 (2003-07-29)
Linum flavum hairy roots were initiated from leaf discs using Agrobacterium rhizogenes strains LBA9402 and TR105 though two other strains, 15834 and A4, were relatively ineffective for induction. Significant variation in coniferin accumulation was observed between hairy root lines originating
Yukiko Tsuji et al.
Journal of agricultural and food chemistry, 52(1), 131-134 (2004-01-08)
To examine the behavior of monolignol and monolignol glucosides in lignin biosynthesis, pentadeutero[9-D(2), 3-OCD(3)]coniferyl alcohol and pentadeutero[9-D(2), 3-OCD(3)]coniferin were synthesized and fed to growing Eucalyptus camaldulensis and Magnolia kobus. The differences in the incorporation patterns of these labeled precursors were
Yukiko Tsuji et al.
Planta, 222(1), 58-69 (2005-06-30)
To gain insight into the behavior of monolignol glucoside in Ginkgo biloba L., we examined glucosides potentially involved in lignin biosynthetic pathway. Coniferin (coniferyl alcohol 4O-beta-D-glucoside) is a strong candidate for the storage form of monolignol. Coniferaldehyde glucoside may also

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service