All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C211H357N67O72
Molecular Weight:
4984.50
UNSPSC Code:
51111800
NACRES:
NA.32
Assay
≥95% (HPLC)
form
lyophilized
composition
Peptide Content, ≥72%
storage condition
protect from light
storage temp.
−20°C
Amino Acid Sequence
Ala-Pro-Leu-Ala-Pro-Arg-Asp-Ala-Gly-Ser-Gln-Arg-Pro-Arg-Lys-Lys-Glu-Asp-Asn-Val-Leu-Val-Glu-Ser-His-Glu-Lys-Ser-Leu-Gly-Glu-Ala-Asp-Lys-Ala-Asp-Val-Asn-Val-Leu-Thr-Lys-Ala-Lys-Ser-Gln
Application
Parathyroid hormone (PTH) is an 84 amino acid hormone involved in calcium regulation, increases calcium levels in blood, in counteraction to calcitonin. PTH also regulates serum phosphate levels and vitamin D biosynthesis. PTH (39-84) and (53-84) are C-terminal peptides of PTH used to resolve PTH activity.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service