Skip to Content
Merck
All Photos(2)

Key Documents

SAE0147

Sigma-Aldrich

Thrombin protease, biotin-tagged

human recombinant,expressed in HEK 293 cells, ≥5000 units/mL

Synonym(s):

Factor IIa, fibrinogenase, thrombase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
NACRES:
NA.77

biological source

human

recombinant

expressed in HEK 293 cells

concentration

≥5000 units/mL

technique(s)

protein extraction: suitable

suitability

suitable for additive or modifier in the separation of proteins or peptides

application(s)

life science and biopharma

shipped in

dry ice

storage temp.

−20°C

General description

Thrombin is an endolytic serine protease that selectively cleaves the Arg–Gly bonds of fibrinogen to form fibrin and release fibrinopeptides A and B.1,2
The optimal cleavage sites for thrombin are as follows:
1. A-B-Pro-Arg-||-X-Y, where A and B are hydrophobic amino acids, and X and Y are nonacidic amino acids.
2. Gly-Arg-||-Gly
Recombinant human thrombin protease is expressed in human HEK 293 cells as a glycoprotein heterodimer. The DTT-reduced protein migrates as two bands of ∼31 kDa (heavy chain) and ∼6 kDa (light chain) on SDS-PAGE. This protein is manufactured in human cells, with no serum. The human cells expression system allows human-like glycosylation and folding, and often supports higher activity of the protein.
This thrombin protease is useful for cleaving recombinant proteins that are expressed as fusion proteins with this sequence between the carrier domain and the protein of interest.

Application

This biotinylated thrombin protease can be used for on-column cleavage of fusion proteins with a thrombin cleavage site. It specifically cleaves the protein of interest from a column-bound fusion protein, leaving the fusion domain or tag bound to the affinity column (e.g., Ni-NTA column) and eluting only the protein of interest.
This method is advantageous over post-elution cleavage for several reasons:
  • It eliminates most impurities normally associated with purification on Ni-chelating columns.
  • It allows gentler elution conditions, with added flexibility in the elution buffer composition. This can mitigate protein aggregation and inactivation.
After cleavage, the protease can be removed with any avidin-conjugated or streptavidin-conjugated beads.

Features and Benefits

The product is supplied in an aqueous buffer of pH 6.0, with 50% (v/v) glycerol.
Catalytic pH range:11.5–10
Optimal pH:11-8.3
(Note: thrombin precipitates at pH ≤5)
Molecular mass:4,12 37.4 kDa
Human isozymes pI range:6.35–7.6
E280 (1%):12 -18.3
This Thrombin protease, biotin-tagged enables on-column cleavage of fusion proteins with a thrombin cleavage site.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service