Skip to Content
Merck
All Photos(3)

Key Documents

SAB5600035

Sigma-Aldrich

Anti-Acetyl-Histone H2B (Lys20) antibody, Rabbit monoclonal

recombinant, expressed in HEK 293 cells, clone RM235, purified immunoglobulin

Synonym(s):

H2BK20ac, Histone H2B (acetyl K20)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

Pricing and availability is not currently available.

recombinant

expressed in HEK 293 cells

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

RM235, monoclonal
recombinant monoclonal

form

buffered aqueous glycerol solution

species reactivity

human

concentration

~1 mg/mL

technique(s)

ELISA: 0.2- 1  μg/mL
immunoblotting: 0.5-2 μg/mL
immunocytochemistry: 1- 2  μg/mL

isotype

IgG

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
53736529589211940
Quality Level

100

Quality Level

200

Quality Level

200

Quality Level

100

form

liquid

form

-

form

-

form

-

bp

169-170 °C/70 mmHg (lit.)

bp

169-170 °C/70 mmHg (lit.)

bp

156-159 °C/10 mmHg (lit.)

bp

-

density

1.076 g/mL at 25 °C (lit.)

density

1.076 g/mL at 25 °C (lit.)

density

1.112 g/mL at 25 °C (lit.)

density

-

mp

−80 °C (lit.)

mp

−80 °C (lit.)

mp

-

mp

54-56 °C (lit.)

Specificity

This antibody reacts to Histone H2B acetylated at Lysine 20 (K20ac). No cross reactivity with non-modified Lysine 20 or other acetylated Lysines in histone H2B.

Immunogen

Synthetic peptide corresponding to Acetyl-Histone H2B (Lys20)

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline containing 50% glycerol, 1% BSA and 0.09% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Need A Sample COA?

This is a sample Certificate of Analysis (COA) and may not represent a recently manufactured lot of this specific product.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service