Skip to Content
Merck
All Photos(2)

Key Documents

SAB5200112

Sigma-Aldrich

Anti-AQP4 antibody produced in rabbit

1 mg/mL, affinity isolated antibody

Synonym(s):

Anti-AQP4, Anti-Aquaporin 4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

mol wt

antigen predicted mol wt 35 kDa

species reactivity

rat, human, mouse

concentration

1 mg/mL

technique(s)

indirect immunofluorescence: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

rat ... Aqp4(25293)

General description

Aquaporin 4 (AQP4) is expressed in the nervous system. The gene ID of the protein is 25293.

Specificity

Detects ~35 kDa. May detect larger glycosylated bands ~50 kDa.

Immunogen

C-terminal peptide

Biochem/physiol Actions

Aquaporin 4 (AQP4) functions as a water channel in the rat brain.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

PBS, pH 7..4, 50% glycerol, and 0.09% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Ceccariglia et al.
Neuroscience, 274, 273-288 (2014-06-10)
Trimethyltin chloride (TMT) is a neurotoxicant producing neuronal degeneration and reactive astrogliosis in the mammalian central nervous system, especially the hippocampus. A previous magnetic resonance imaging investigation in TMT-treated rats evidenced dilation of lateral ventricles, also suggesting alterations in blood-brain
Teruyuki Ishikura et al.
Journal of neuroinflammation, 18(1), 181-181 (2021-08-23)
Intractable neuropathic pain is a common symptom of neuromyelitis optica spectrum disorder (NMOSD). However, the underlying mechanism of NMOSD pain remains to be elucidated. In this study, we focused on ATP, which is one of the damage-associated molecular patterns, and
Elisa Vizzaccaro et al.
Muscle & nerve, 57(5), 856-858 (2017-12-02)
Aquaporins (AQPs) are a family of transmembrane proteins involved in the maintenance of osmotic gradients. AQP4 is abundant in skeletal muscle, where it seems to be associated with glycolytic metabolism. We investigated the pattern of expression of AQP4 in normal
Alex J Smith et al.
Acta neuropathologica communications, 7(1), 74-74 (2019-05-10)
Redistribution of the water channel aquaporin-4 (AQP4) away from astrocyte endfeet and into parenchymal processes is a striking histological feature in mouse models of Alzheimer's disease (AD) and other neurological conditions with prominent astrogliosis. AQP4 redistribution has been proposed to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service