Skip to Content
Merck
All Photos(1)

Key Documents

SAB4700284

Sigma-Aldrich

Monoclonal Anti-CD15 antibody produced in mouse

clone MEM-158, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-FUT4, Anti-Lewis x Blood Group antigen

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

MEM-158, monoclonal

form

buffered aqueous solution

species reactivity

human

concentration

1 mg/mL

technique(s)

flow cytometry: suitable

isotype

IgM

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... FUT4(2526)

General description

Cluster of differentiation 15 (CD15), also known as fucosyltransferase 4 (FUT4)/ Lewis x blood group antigen, is encoded by the gene mapped to long arm of human chromosome 11q21. It is ubiquitously expressed in various tissues such as gastrointestinal tract, brain and leukocytes.
The antibody MEM-158 reacts with CD15, a cell membrane molecule 3-fucosyl-N-acetyllactosamine (3-FAL) strongly expressed on granulocytes, monocytes, macrophages, mast cells; it is also present on Langerhans cells and some myeloid precursors cells.

Immunogen

Human granulocytes

Application

The reagent is designed for Flow Cytometry analysis using 10 μL reagent / 100 μL of sample.

Biochem/physiol Actions

Cluster of differentiation 15 (CD15)/fucosyltransferase 4 catalyzes the synthesis of Lex on the surface of the proliferating cells, in organ buds progressing in mesenchyma, during human embryogenesis. Overexpression of the gene has been associated with development of various type of cancers such as colon cancer, pancreatic cancer, gastric cancer and lung adenocarcinoma. siRNAs of FUT1/4 can be used as a potent therapeutic agent in human LeY-expressing cancers.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4, with 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sakiko Masuda et al.
The American journal of pathology, 189(4), 839-846 (2019-01-25)
Antineutrophil cytoplasmic antibody (ANCA)-associated vasculitis (AAV) is characterized by the production of ANCAs and systemic necrotizing vasculitis in small vessels. Disordered regulation of neutrophil extracellular traps (NETs) is critically involved in the pathogenesis of AAV. NETs are web-like DNA decorated
The gene encoding myeloid alpha-3-fucosyl-transferase (FUT4) is located between D1 1S388 and D11S919 on 11q21.
Reguigne I
Cytogenetics and Cell Genetics, 66, 104-106 (1994)
Wei Jin et al.
Annals of surgical oncology, 26(2), 635-643 (2018-10-31)
Tumor-infiltrating neutrophils (TINs) indicate poor prognosis for patients with pancreatic ductal adenocarcinoma (PDAC). Activated neutrophils can generate neutrophil extracellular traps (NETs). Little is known about the presence and prognostic significance of tumor-infiltrating NETs in PDAC. This study enrolled 317 patients
Miaomiao Gao et al.
Journal of cellular and molecular medicine, 23(3), 2032-2041 (2018-12-27)
Alcoholic liver disease (ALD) is a progressive liver disease that can cause a series of complications, including cirrhosis, liver failure and hepatocellular carcinoma. Granulocytic myeloid-derived suppressor cell (gMDSC) populations have been observed to expand in various liver diseases and to
Filippo Veglia et al.
Nature, 569(7754), 73-78 (2019-04-19)
Polymorphonuclear myeloid-derived suppressor cells (PMN-MDSCs) are pathologically activated neutrophils that are crucial for the regulation of immune responses in cancer. These cells contribute to the failure of cancer therapies and are associated with poor clinical outcomes. Despite recent advances in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service