Skip to Content
Merck
All Photos(1)

Key Documents

SAB4600436

Sigma-Aldrich

Anti-Human IgM (μ chain), CF647 antibody produced in goat

~2 mg/mL, affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46
clone:
polyclonal
application:
ELISA (i)
FACS
ICC
IF
IHC
WB
species reactivity:
human
technique(s):
flow cytometry: 1-10 μg/mL
immunocytochemistry: suitable
immunohistochemistry: suitable
indirect ELISA: suitable
indirect immunofluorescence: 1-10 μg/mL
western blot: suitable
citations:
4

biological source

goat

Quality Level

conjugate

CF 647 conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

concentration

~2 mg/mL

technique(s)

flow cytometry: 1-10 μg/mL
immunocytochemistry: suitable
immunohistochemistry: suitable
indirect ELISA: suitable
indirect immunofluorescence: 1-10 μg/mL
western blot: suitable

fluorescence

λex 650 nm; λem 665 nm

storage temp.

−20°C

General description

CF647 goat anti—human IgM (μ chain) is labeled with our bright, deep red-fluorescent CF647 dye and is prepared from antibodies purified by human IgM—affinity chromatography. The antibodies react specifically with IgM heavy chains (μ chains) and not with immunoglobulin light chains. To minimize cross-reactivity, it has been adsorbed human IgG prior to conjugation
During the B-cell development, IgM (immunoglobulin M) is the first expressed antibody present as a monomer on the surface of naive B cells. Antigenic stimulation and upon maturation, the monomeric IgM units (190kDa), join together to form a pentameric immunoglobulin structure. It also but rarely forms a hexameric structure. The pentameric IgM is linked by a disulfide bond, a polypeptide chain and a J-chain. The μ-chain consists of five domains- VH, Cμ1, Cμ2, Cμ3 and Cμ4.

Specificity

Binds human IgM heavy chain (mu chain) (minimal cross-reactivity IgG and IgA heavy chains, or immuglobulin light chains).

Immunogen

Human IgM(H+L)

Biochem/physiol Actions

The J-chain of the multimeric IgM (immunoglobulin M) structure is responsible for secretion at the mucosal surfaces. The pentameric form of IgM, has a greater affinity towards antigens (with multiple repeating epitopes) compared to the monomers. The pentameric IgM enables better opsonization of antigens for destruction and fixing complement. IgM is essential of the activation of classical complement system. Reduced serum IgM levels is observed in Bruton′s agammaglobulinemia an X-linked disorder and transient hypogammaglobulinemia in infants.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied in phosphate buffered saline with 0.05% sodium azide, 50% glycerol and 0.2% BSA.

Preparation Note

Protect from light. The antibody solution should be gently mixed before use.

Legal Information

This product is distributed by Sigma-Aldrich Co. under the authorization of Biotium, Inc. This product is covered by one or more US patents and corresponding patent claims outside the US patents or pending applications owned or licensed by Biotium, Inc. including without limitation: 12/334,387; 12/607,915; 12/699,778; 12/850,578; 61/454,484. In consideration of the purchase price paid by the buyer, the buyer is hereby granted a limited, non-exclusive, non-transferable license to use only the purchased amount of the product solely for the buyer′s own internal research in a manner consistent with the accompanying product literature. Except as expressly granted herein, the sale of this product does not grant to or convey upon the buyer any license, expressly, by implication or estoppel, under any patent right or other intellectual property right of Biotium, Inc. Buyer shall not resell or transfer this product to any third party, or use the product for any commercial purposes, including without limitation, any diagnostic, therapeutic or prophylactic uses. This product is for research use only. Any other uses, including diagnostic uses, require a separate license from Biotium, Inc. For information on purchasing a license to use this product for purposes other than research, contact Biotium, Inc., 3159 Corporate Place, Hayward, CA 94545, Tel: (510) 265-1027. Fax: (510) 265-1352. Email: btinfo@biotium.com.
CF is a trademark of Biotium, Inc.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Handbook of Human Immunology, 74-74 (2008)
Immunobiology: The Immune System in Health and Disease (2005)
Serum Immunoglobulin M Concentration Is Positively Related to Metabolic Syndrome in an Adult Population: Tianjin Chronic Low-Grade Systemic Inflammation and Health (TCLSIH) Cohort Study
Kun Song
PLoS ONE, 9(2), e88701-e88701 (2014)
Structure and function of immunoglobulins.
Schroeder Jr H W and Cavacini L
The Journal of Allergy and Clinical Immunology, 125(2), S41-S52 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service