Skip to Content
Merck
All Photos(6)

Key Documents

SAB4600052

Sigma-Aldrich

Anti-Chicken IgY (H+L), CF 488A antibody produced in rabbit

~2 mg/mL, affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

CF 488A conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

chicken

concentration

~2 mg/mL

technique(s)

flow cytometry: 1-10 μg/mL
immunocytochemistry: suitable
immunohistochemistry: suitable
indirect immunofluorescence: 1-10 μg/mL

fluorescence

λex 490 nm; λem 515 nm

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Immunoglobulin Y is one of the antibodies produced in birds. It is generally related to mammalian IgG. However, IgY has different characteristics compared to mammalian IgG, for example, IgY does not bind with mammalian complements nor Rheumatoid factor, which are present in mammalian serum and anti-mammalian IgG antibodies, which often cross-react to IgG from different mammalian species, exhibit low cross-reactivity to IgY. These characteristics of IgY is beneficial when it is used as a biochemical/clinical reagent. Immunoglobulin Y, present in chicken blood serum, has the capability to pass across the follicular epithelium of the ovary and aggregate in the yolk during oogenesis. Structurally IgY and mammalian IgG differ in the heavy chain with the molecular weight of 68,000 and 50,000 respectively. Mammalian IgG has slightly higher stability compared to IgY.
CF488A is a green fluorescent dye with excitation argon laser line at 488 nm. CF488A is minimally charged, thus it reduces non-specific binding of antibody conjugates. CF488A is a highly water-soluble and pH-insensitive dye with molecular weight of ~710. This dye has absorption/emission maxima at 490/515 nm and it is highly photostable.

Specificity

Binds all chicken IgYs

Immunogen

chicken IgY (H+L)

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied in phosphate buffered saline with 0.05% sodium azide, 50% glycerol and 2 mg/mL bovine serum albumin.

Preparation Note

Protect from light.

Legal Information

This product is distributed by Sigma-Aldrich Co. under the authorization of Biotium, Inc. This product is covered by one or more US patents and corresponding patent claims outside the US patents or pending applications owned or licensed by Biotium, Inc. including without limitation: 12/334,387; 12/607,915; 12/699,778; 12/850,578; 61/454,484. In consideration of the purchase price paid by the buyer, the buyer is hereby granted a limited, non-exclusive, non-transferable license to use only the purchased amount of the product solely for the buyer′s own internal research in a manner consistent with the accompanying product literature. Except as expressly granted herein, the sale of this product does not grant to or convey upon the buyer any license, expressly, by implication or estoppel, under any patent right or other intellectual property right of Biotium, Inc. Buyer shall not resell or transfer this product to any third party, or use the product for any commercial purposes, including without limitation, any diagnostic, therapeutic or prophylactic uses. This product is for research use only. Any other uses, including diagnostic uses, require a separate license from Biotium, Inc. For information on purchasing a license to use this product for purposes other than research, contact Biotium, Inc., 3159 Corporate Place, Hayward, CA 94545, Tel: (510) 265-1027. Fax: (510) 265-1352. Email: btinfo@biotium.com.
CF is a trademark of Biotium, Inc.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Immunoglobulins of the non-galliform birds: antibody expression and repertoire in the duck.
Lundqvist ML
Developmental and Comparative Immunology, 30(1-2), 93-100 (2006)
Molecular stability of chicken and rabbit immunoglobulin G.
Shimizu M
Bioscience, Biotechnology, and Biochemistry, 56(2), 270-274 (1992)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service