Skip to Content
Merck
All Photos(6)

Key Documents

SAB4504403

Sigma-Aldrich

Anti-phospho-FAK (pTyr397) antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 119 kDa

species reactivity

rat, mouse, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:5000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pTyr397)

Gene Information

human ... PTK2(5747)

General description

Protein tyrosine kinase 2 (PTK2), also known as FAK (focal adhesion kinase), is encoded by the gene mapped to human chromosome 8q24-qter. The encoded protein is localized to cell adhesion sites and extracellular matrix.

Immunogen

The antiserum was produced against synthesized peptide derived from human FAK around the phosphorylation site of Tyr397.

Immunogen Range: 364-413

Biochem/physiol Actions

Protein tyrosine kinase 2 (PTK2) is a non-receptor tyrosine kinase which is known to regulate cell migration, proliferation, cell survival and invasiveness. Increased expression of FAK (focal adhesion kinase) leads to metastatic breast tumors and lung cancer. Inhibition of FAK tyrosine kinase activity stops various cellular process necessary for tumor growth. Hence, FAK inhibitors are considered to be potential therapeutic methods for cancer treatment. FAK is one of the important components of integrin adhesion complexes (IACs)and it plays a vital role in maintaining IAC dynamics. Cell interactions with components of extracellular matrix and a number of agonists, such as neuropeptides, activate Ptk2. Ptk2, mediates cell signaling through its kinase dependent/independent functions. Upregulation of the gene is observed in hepatocellular carcinoma, which is accompanied with portal venous invasion and intrahepatic metastasis.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F T Fiedorek et al.
Mammalian genome : official journal of the International Mammalian Genome Society, 6(2), 123-126 (1995-02-01)
Focal adhesion kinase (pp125FAK or FAK) is a cytoplasmic protein-tyrosine kinase stimulated in response to cell interactions with extracellular matrix components and by exposure to a variety of agonists, including neuropeptides. FAK lacks Src-homology SH2 and SH3 domains, is highly
T Konry et al.
Talanta, 75(2), 564-571 (2008-03-29)
We present herein an effective and versatile method to fabricate a micro-patterned structure of conductive polymer, poly(pyrrole-benzophenone), on Indium Tin Oxide (ITO) glass chips for the subsequent photo-immobilization of various bioreceptor, antigens. Such methodologies are based on photolithography of ITO
Focal adhesion kinase inhibitors in combination with erlotinib demonstrate enhanced anti-tumor activity in non-small cell lung cancer.
Howe G A, et al.
PLoS ONE, 11(3), e0150567-e0150567 (2016)
Iliet Payan et al.
International journal of oncology, 48(5), 2043-2054 (2016-03-18)
Focal adhesion kinase (FAK) is a non-receptor tyrosine kinase known to promote cell migration and invasiveness. Overexpression and increased activity of FAK are closely associated with metastatic breast tumors and are linked to poor prognosis. This study discovered an inverse
Focal adhesion kinase depletion reduces human hepatocellular carcinoma growth by repressing enhancer of zeste homolog 2.
Gnani D, et al.
Cell Death and Differentiation, 24(5), 889-889 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service