Skip to Content
Merck
All Photos(3)

Key Documents

SAB4503583

Sigma-Aldrich

Anti-CD4 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

CD4, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4 precursor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 51 kDa

species reactivity

human, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:1000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CD4(920)

General description

Anti-CD4 Antibody detects endogenous levels of total CD4 protein.
CD4 (CD4 molecule) is present in the surface of specific leukocytes. CD4 contains four ecto-domains (D1-D4) of which D1, D2, and D4 contain disulfide bonds. It is mapped to human chromosome 12p13.31.

Immunogen

The antiserum was produced against synthesized peptide derived from human CD4.

Immunogen Range: 401-450

Application

Anti-CD4, C-Terminal antibody has been used in immunohistochemical staining.

Biochem/physiol Actions

CD4 (CD4 molecule) helps in the activation of immunostimulatory T-cells. It serve as a primary receptor for HIV-1 entry.This molecule plays an important role on cell membrane mechanical characteristics and can be used in the development of cell biology and medicine.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

M I Ashraf et al.
American journal of transplantation : official journal of the American Society of Transplantation and the American Society of Transplant Surgeons, 16(3), 808-820 (2015-11-26)
Lipocalin 2 (Lcn2) is rapidly produced by damaged nephron epithelia and is one of the most promising new markers of renal injury, delayed graft function and acute allograft rejection (AR); however, the functional importance of Lcn2 in renal transplantation is
Exogenous Lipocalin 2 Ameliorates Acute Rejection in a Mouse Model of Renal Transplantation
Ashraf MI, et al.
American Journal of Transplantation : Official Journal of the American Society of Transplantation and the American Society of Transplant Surgeons, 16(3), 808-820 (2016)
Fang Zhang et al.
Frontiers in immunology, 10, 2392-2392 (2019-11-05)
Background: Stroke patients with diabetes suffer from higher mortality rate and worsened neurological outcome. However, the responses of immune system to cerebral ischemia in the setting of diabetes remain poorly understood. Methods: In this study, we investigated the temporal profile
Human CD4 Metastability Is a Function of the Allosteric Disulfide Bond in Domain 2
Owen GR, et al.
Biochemistry, 55(15), 2227-2237 (2016)
Isochromosomes 12p and 9p: parental origin and possible mechanisms of formation
Dutly F, et al.
European Journal of Human Genetics, 6(2), 140-144 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service