Skip to Content
Merck
All Photos(3)

Key Documents

SAB4503499

Sigma-Aldrich

Anti-Cyclin A antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Cyclin A1, cyclin A1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 52 kDa

species reactivity

rat, human, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:10000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CCNA1(8900)

Related Categories

General description

Anti-Cyclin A Antibody detects endogenous levels of total Cyclin A protein.
CCNA1 (cyclin A1) is a member of the highly conserved cyclin family. There are two A-type cyclins, namely cyclin A1 and cyclin A2. Cyclin A1 is expressed at high levels in testis and expressed at low levels in most other tissues. This gene is mapped to human chromosome 13q13.

Immunogen

The antiserum was produced against synthesized peptide derived from human Cyclin A.

Immunogen Range: 221-270

Application

Anti-Cyclin A antibody has been used in western blotting.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Biochem/physiol Actions

CCNA1 (cyclin A1) controls cell apoptosis. Cyclin A2 is required for DNA replication and proliferation in somatic cells. CCNA1 plays an important role in head and neck carcinoma and cervical and nasopharyngeal cancers.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Correlation of CCNA1 promoter methylation with malignant tumors: a meta-analysis introduction.
Yang B, et al.
BioMed Research International, 2015, 134027-134027 (2015)
Cyclin A1 shows age-related expression in benign tonsils, HPV16-dependent overexpression in HNSCC and predicts lower recurrence rate in HNSCC independently of HPV16.
Weiss D, et al.
BMC Cancer, 12(1), 259-259 (2012)
Xiaomin Su et al.
BMC developmental biology, 20(1), 22-22 (2020-11-19)
Tissue regeneration mediated by mesenchymal stem cells (MSCs) is deemed a desirable way to repair teeth and craniomaxillofacial tissue defects. Nevertheless, the molecular mechanisms about cell proliferation and committed differentiation of MSCs remain obscure. Previous researches have proved that lysine
MiR-129-5p Inhibits Proliferation and Invasion of Chondrosarcoma Cells by Regulating SOX4/Wnt/β-Catenin Signaling Pathway.
Zhang P, et al.
Cellular Physiology and Biochemistry, 42(1), 242-253 (2017)
Depletion of HOXA5 inhibits the osteogenic differentiation and proliferation potential of stem cells from the apical papilla.
Li W, et al.
Cell Biology International, 42(1), 45-52 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service