Skip to Content
Merck
All Photos(3)

Key Documents

SAB4503250

Sigma-Aldrich

Anti-CAMK2 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

CAMK2A, CaM-kinase II α chain, CaMK-II α subunit, CaMKII-α, Calcium/calmodulin-dependent protein kinase type II α chain

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 54 kDa

species reactivity

rat, mouse, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:10000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CAMK2A(815)

General description

Anti-CAMK2 Antibody detects endogenous levels of total CAMK2 protein.
Calcium/calmodulin dependent protein kinase II α (CAMK2) is a calcium-activated serine/threonine kinase. The gene is located on human chromosome 5q32. CAMK2 is mainly expressed in the central nervous system. It consists of a kinase domain, a regulatory domain and an association domain.

Immunogen

The antiserum was produced against synthesized peptide derived from human CaMK2 alpha/delta.

Immunogen Range: 256-305

Biochem/physiol Actions

Calcium/calmodulin dependent protein kinase II α (CAMK2) is essential for long-term potentiation (LTP) and spatial memory. It regulates synaptic plasticity. Dysfunction in this gene is associated with schizophrenia, epilepsy and autism spectrum disorders.
Calmodulin dependent protein kinase II complex δ (CAMK2D) modulates the functions of ion channels and Ca2+ handling proteins. It also regulates the phosphorylation and acetylation of histones. It has been shown to be overexpressed in the myocardium of diabetic patients.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

De novo variants in CAMK 2A and CAMK 2B cause neurodevelopmental disorders
Akita T, et al.
Annals of Clinical and Translational Neurology, 5(3), 280-296 (2018)
A homozygous loss-of-function CAMK2A mutation causes growth delay, frequent seizures and severe intellectual disability
Chia PH, et al.
eLife, 7, e32451-e32451 (2018)
Targeted multiplexed selected reaction monitoring analysis evaluates protein expression changes of molecular risk factors for major psychiatric disorders
Wesseling H, et al.
The International Journal of Neuropsychopharmacology, 18(1) (2015)
Osiris Germán Idelfonso-García et al.
Antioxidants (Basel, Switzerland), 13(3) (2024-03-28)
Aging is characterized by increased reactive species, leading to redox imbalance, oxidative damage, and senescence. The adverse effects of alcohol consumption potentiate aging-associated alterations, promoting several diseases, including liver diseases. Nucleoredoxin (NXN) is a redox-sensitive enzyme that targets reactive oxygen
Matilda K Dahlqvist et al.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism, 40(10), 2010-2025 (2019-10-28)
Gamma activity arising from the interplay between pyramidal neurons and fast-spiking parvalbumin (PV) interneurons is an integral part of higher cognitive functions and is assumed to contribute significantly to brain metabolic responses. Cerebral metabolic rate of oxygen (CMRO2) responses were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service