Skip to Content
Merck
All Photos(1)

Key Documents

SAB4502201

Sigma-Aldrich

Anti-PLAGL1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

LOT-1, Lost on transformation 1, Pleiomorphic adenoma-like protein 1, Tumor supressor ZAC, Zinc finger protein PLAGL1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 50 kDa

species reactivity

human

concentration

~1 mg/mL

technique(s)

ELISA: 1:10000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PLAGL1(5325)

General description

The gene PLAGL1 (pleiomorphic adenoma-like protein 1), PLAGL2 and PLAG1 form a subfamily of PLAG family of zinc finger proteins that recognize DNA and RNA. The three members share similarity at the amino-terminal, but are functionally different, varying in their DNA binding capacities. The gene is mapped to human chromosome 6q24.2, a region implicated in several cancers.

Immunogen

The antiserum was produced against synthesized peptide derived from human PLAGL1.

Immunogen Range: 311-360

Biochem/physiol Actions

The maternally-imprinted gene, PLAGL1 (pleiomorphic adenoma-like protein 1), functions as an oncogene as well as a tumor suppressor depending on the cell context. It has been found to exhibit anti-proliferative properties and regulate apoptosis and cell-cycle arrest via p53.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The tumorigenic diversity of the three PLAG family members is associated with different DNA binding capacities.
Hensen K
Cancer Research, 62, 1510-1517 (2002)
Anne-Lise Peille et al.
PloS one, 8(11), e80741-e80741 (2013-11-22)
Soft tissue sarcomas (STS) are rare, complex tumors with a poor prognosis. The identification of new prognostic biomarkers is needed to improve patient management. Our aim was to determine the methylation status of the 118 CpG sites in the PLAGL1
Transcriptional activation capacity of the novel PLAG family of zinc finger proteins.
Kas K
The Journal of Biological Chemistry, 273, 23026-23032 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service