Skip to Content
Merck
All Photos(1)

Key Documents

SAB4502023

Sigma-Aldrich

Anti-NOTCH4 (Cleaved-Val1432), C-Terminal antibody produced in rabbit

affinity isolated antibody

Synonym(s):

INT3, NOTC4, NOTCH3, neurogenic locus notch 4, notch 4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 59 kDa

species reactivity

human

concentration

~1 mg/mL

technique(s)

ELISA: 1:1000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

proteolytically cleaved (Val1432)

Gene Information

human ... NOTCH4(4855)

General description

Anti-NOTCH4 (Cleaved-Val1432) antibody detects endogenous levels of fragment of activated NOTCH4 (Cleaved-Val1432) protein.
The NOTCH4 gene is mapped to human chromosome 6p21.32.

Immunogen

The antiserum was produced against synthesized peptide derived from human NOTCH4.

Immunogen Range: 1401-1450

Biochem/physiol Actions

The NOTCH4 gene is associated with the process of angiogenesis and homeostasis during developmental process. NOTCH4 plays a role in Notch signaling pathway which mediates epithelial cell and stem cell homeostasis. Notch signaling pathway is induced in a number of cancer types including colon, breast, pancreatic, cervical, head and neck, renal carcinoma, prostate cancer, Large-cell and Hodgkin lymphomas.Thus, NOTCH4 is known to participate in tumorigenesis. This gene is involved in the development of central nervous system and controls neuronal maturation. Mutations in the gene might be associated with schizophrenia.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seema R Lalani et al.
BMC medical genetics, 6, 8-8 (2005-02-16)
CHARGE syndrome is a complex of birth defects including coloboma, choanal atresia, ear malformations and deafness, cardiac defects, and growth delay. We have previously hypothesized that CHARGE syndrome could be caused by unidentified genomic microdeletion, but no such deletion was
Harishankar Mk et al.
Life sciences, 156, 38-46 (2016-05-20)
Despite the development of several therapeutic strategies in the past decades, clinicians have failed to improve the survival rate of oral squamous cell carcinoma patients due to the highly metastatic nature of the disease and its high recurrence rate. However
Jianwen Xiong et al.
Oncotarget, 8(8), 13157-13165 (2017-01-07)
Notch signalling is aberrantly activated in human non-small cell lung cancer (NSCLC). Nevertheless, the prognostic roles of mRNA expression of four Notch receptors in NSCLC patients remain elusive. In this report, we reported the prognostic roles of Notch receptors in
Bao Zhang et al.
BioMed research international, 2015, 408096-408096 (2015-11-26)
NOTCH4 regulates signaling pathways associated with neuronal maturation, a process involved in the development and patterning of the central nervous system. The NOTCH4 gene has also been identified as a possible susceptibility gene for schizophrenia (SCZ). The objective of this
Xiaoxia Liu et al.
Reproduction (Cambridge, England), 152(1), 47-55 (2016-04-14)
Preeclampsia is a serious complication of pregnancy and is closely related to endothelial dysfunction, which can be repaired by endothelial progenitor cells (EPCs). The DLL4/NOTCH-EFNB2 (ephrinB2) cascade may be involved in the pathogenesis of preeclampsia by inhibiting the biological activity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service