Skip to Content
Merck
All Photos(2)

Key Documents

SAB4501651

Sigma-Aldrich

Anti-Keratin 5 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

58 kDa cytokeratin, CK-5, Cytokeratin-5, K5, Keratin type II cytoskeletal 5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 62 kDa

species reactivity

mouse, human, rat

concentration

~1 mg/mL

technique(s)

ELISA: 1:10000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KRT5(3852)

General description

Anti-Keratin 5 Antibody detects endogenous levels of total Keratin 5 protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human Keratin 5.

Immunogen Range: 541-590

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Verena Sigl et al.
Cell research, 26(7), 761-774 (2016-06-01)
Breast cancer is the most common female cancer, affecting approximately one in eight women during their life-time. Besides environmental triggers and hormones, inherited mutations in the breast cancer 1 (BRCA1) or BRCA2 genes markedly increase the risk for the development
Meixia Che et al.
Nature communications, 12(1), 6377-6377 (2021-11-06)
Endocrine therapies for prostate cancer inhibit the androgen receptor (AR) transcription factor. In most cases, AR activity resumes during therapy and drives progression to castration-resistant prostate cancer (CRPC). However, therapy can also promote lineage plasticity and select for AR-independent phenotypes
Marc Fernandez-Guerrero et al.
Proceedings of the National Academy of Sciences of the United States of America, 117(48), 30509-30519 (2020-11-18)
Vertebrate Hox genes are critical for the establishment of structures during the development of the main body axis. Subsequently, they play important roles either in organizing secondary axial structures such as the appendages, or during homeostasis in postnatal stages and
Shannon Conway et al.
JID innovations : skin science from molecules to population health, 4(4), 100283-100283 (2024-06-03)
The skin is a multifunctional organ, forming a barrier between the external and internal environment, thereby functioning as a safeguard against extrinsic factors. Autophagy has been implicated in epidermal differentiation and in preserving skin homeostasis. LC3-associated phagocytosis (LAP) uses some
Norio Kitagawa et al.
Anatomical science international, 94(2), 163-171 (2018-10-26)
Nisin is a food preservative produced by Lactococcus lactis subsp. lactis. Previous blood biochemical research revealed that nisin has physiological effects in mammals; although the site of action has yet to be identified, keratinocytes have been proposed as a possible

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service