Skip to Content
Merck
All Photos(2)

Key Documents

SAB4500671

Sigma-Aldrich

Anti-DRD1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

D(1A) dopamine receptor, DRD1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 49 kDa

species reactivity

mouse, rat, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:5000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DRD1(1812)

General description

Anti-DRD1 Antibody detects endogenous levels of total DRD1 protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human DRD1.

Immunogen Range: 135-184

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mireia Casanovas et al.
Addiction biology, 26(5), e13017-e13017 (2021-02-10)
Cocaine not only increases brain dopamine levels but also activates the sigma1 receptor (σ1 R) that in turn regulates orexigenic receptor function. Identification of interactions involving dopamine D1 (D1 R), ghrelin (GHS-R1a ), and σ1 receptors have been addressed by
Pablo Garrido-Gil et al.
Molecular neurobiology, 55(9), 7297-7316 (2018-02-07)
The exact mechanism of gut dysfunction in Parkinson's disease and, conversely, the role of gut pathology in brain dopaminergic degeneration are controversial. We investigated the effects of nigral lesions on the colonic neurotransmission, the effect of gut inflammation on the
Pablo Garrido-Gil et al.
Oncotarget, 9(13), 10834-10846 (2018-03-16)
Gastrointestinal dysfunction is a common problem in the elderly. Aging-related changes in interactions between local dopaminergic and renin-angiotensin systems (RAS) have been observed in the brain, renal and vascular tissues. However, it is not known if these interactions also occur

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service