Skip to Content
Merck
All Photos(1)

Documents

SAB4500488

Sigma-Aldrich

Anti-PARP (Cleaved-Gly215) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

ADP-ribosyltransferase 1, ADPRT, ADPRT1, PARP, pADPRT-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 89 kDa

species reactivity

human

concentration

~1 mg/mL

technique(s)

ELISA: 1:1000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

proteolytically cleaved (Gly215)

Gene Information

human ... PARP1(142)

Related Categories

General description

Anti-PARP (Cleaved-Gly215) Antibody detects endogenous levels of fragment of activated PARP (Cleaved-Gly215) protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human PARP.

Immunogen Range: 196-245

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lijun Wang et al.
The journal of gene medicine, 22(12), e3254-e3254 (2020-07-18)
Non-small cell lung cancer (NSCLC) is a lethal tumor resulting in a large number of cancer-related deaths globally. Long noncoding RNAs (lncRNAs) may modulate tumor initiation and metastasis. Although dysregulation of lncRNA cancer susceptibility 19 (CASC19) is validated in NSCLC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service