Skip to Content
Merck
All Photos(1)

Key Documents

SAB4200861

Sigma-Aldrich

Anti-PSTAIR antibody, Mouse monoclonal

clone PSTAIR, purified from hybridoma cell culture

Synonym(s):

pan-CDKs

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

antibody form

purified from hybridoma cell culture

Quality Level

clone

PSTAIR

form

liquid

species reactivity

rat, monkey, human, mouse

concentration

~1 mg/mL

technique(s)

immunoblotting: 0.5-1.0 μg/mL using whole extracts of COS7 cells.

isotype

IgG1

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Cell division is a fundamental biological process, consisting of the splitting of the cell and its genetic material into two daughter cells. Mitosis is a cell division process that results in the formation of two new nuclei, each having the same number of chromosomes as the parental nucleus. During the cell cycle of most somatic cells, DNA synthesis (S-phase) and mitosis (M-phase) are separated by two "growth" stages (G1 and G2) of varying duration. A typical eukaryotic cell sequentially passes through G1, S, G 2, and M and back into G1 during each single cycle.9 Maturation-promoting factor (MPF), originally identified during meiosis in frog oocytes, is a cytoplasmic factor, which is highly conserved among a wide range of species. Itplays a key role in the progression of the cell cycle from interphase (G2) to metaphase (M), in both meiosis and mitosis.

Specificity

Monoclonal Anti-PSTAIR antibody specifically recognizes the evolutionary conserved PSTAIR sequence. The antibody recognizes 31-34 kDa proteins (1-4 bands) in immunoblotting.1-7 Cross-reactivity has been observed with human, monkey, mouse,1,8 Japanese quail,1,3 fish (goldfish,4-7 carp,1,5,7 eel,1 starfish,1 and amago salmon1 ), Xenopus,1 cliliates,1 and plants (lily,1,2 and onion1 ).

Application

The antibody may be used for Immunoblotting based assays.

Biochem/physiol Actions

The cell cycle can be considered as a cyclin-dependent kinases (cDKs) cycle, which is controlled by biochemical modifications such as phosphorylation of CDKs and formation of complex(es) with other proteins, including the cyclins.10 CDKs are key regulators of cell cycle progression.11,12 and are closely related in size (35-40 kDa) and sequence (>40% identity). Each CDK interacts with a specific subset of cyclins which activate them by enabling their phosphorylation at specific residues.

In the fission yeast Shizosaccharomyces pombe, a single major CDK has been identified (cdc2) while in human cells, the growing list of CDKs now includes also cdc2 (Cdk1), and Cdk2 to Cdk7. The typical CDK catalytic subunit contains a 300 amino acid catalytic core that is completely inactive in monomeric and unphosphorylated state.13 15.14 CDKs are constitutively expressed throughout the cell cycle and are activated and inactivated by specific kinases and phosphatases.15,16 In every eukaryote examined, CDKs contain an evolutionary conserved 16 amino acid sequence called PSTAIR (EGVPSTAIREISLLKE) which distinguishes them from other protein kinases. The PSTAIR motif is involved in the complex formation with cyclins. The availability of monoclonal antibody reacting specifically with the PSTAIR sequence1,17 enables the subcellular detection and localization of the various CDKs and examination of substrate interactions, in a variety of organisms.

Physical form

Supplied as a solution in 0.01 M phosphate buffered saline pH 7.4, containing 15 mM sodium azide as a preservative.

Storage and Stability

For continuous use, store at 2-8°C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog  our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service