Skip to Content
Merck
All Photos(3)

Documents

SAB4200416

Sigma-Aldrich

Anti-Tensin 3 antibody, Mouse monoclonal

clone TN-17, purified from hybridoma cell culture

Synonym(s):

Monoclonal Anti-TEM6, Monoclonal Anti-TENS1, Monoclonal Anti-TNS3, Monoclonal Anti-Tensin 3 antibody produced in mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

TN-17, monoclonal

form

buffered aqueous solution

mol wt

antigen ~170 kDa

species reactivity

human

concentration

~1.0 mg/mL

technique(s)

indirect immunofluorescence: 2.5-5.0 μg/mL using PC3 cells.
western blot: 0.5-1.0 μg/mL using extracts of SW-620 cells.

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TNS3(64759)

General description

Monoclonal Anti-Tensin 3 (mouse IgG1 isotype) is derived from the hybridoma TN-17 produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with a synthetic peptide corresponding to an internal sequence of human Tensin 3. Tensins are multidomain proteins that are characterized with homologous C1, protein phosphatases (PTPases) , C2, Src homology 2 (SH2) domains and pTyr-binding (PTB) domains.
Tensin 3 (TNS3) is encoded by the gene mapped to human chromosome 7p12.3. TNS3 transcript is expressed at high level in thyroid and at moderate level in placenta. In addition, the protein is also expressed at minimal levels in brain, heart, lung, liver, pancreas, kidney, skeletal muscle, white blood cells and prostate. TNS3 is a member of tensin protein family.

Immunogen

synthetic peptide corresponding to an internal sequence of human Tensin 3, conjugated to KLH. The isotype is determined using a double diffusion immunoassay using Mouse Monoclonal Antibody Isotyping Reagents (Sigma ISO-2).

Application

Monoclonal Anti-Tensin 3 antibody produced in mouse has also been used in immunofluorescence.
Monoclonal Anti-Tensin 3 antibody produced in mouse has been used in western blotting.

Biochem/physiol Actions

Tensin 3 (TNS3) plays a vital role in stimulation of epidermal growth factor (EGF) signaling pathway. The encoded protein supports fundamental role of thyroid hormone in normal thyroid tissues. Decreased expression of the gene leads to development of human renal cell carcinoma (RCC).
Tensin 3 facilitates cell migration and tumorigenesis in cell lines from advanced lung cancer, breast cancer and melanoma. In these cell lines, The protein is phosphorylated by Src in its SH2 domain, leading to tumorigenesis and metastasis, implicating tensin 3 as an oncoprotein regulated by Src.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

CpG dinucleotide-specific hypermethylation of the TNS3 gene promoter in human renal cell carcinoma
Carter JA
Epigenetics, 8, 739-747 (2013)
Increased expression levels of tensin3 mRNA in thyroid functional adenomas as compared to non-functioning adenomas.
Maeda I
Experimental and Clinical Endocrinology & Diabetes, 117, 191-193 (2009)
Musashi-1 Enhances Glioblastoma Cell Migration and Cytoskeletal Dynamics through Translational Inhibition of Tensin3
Chen HY
Scientific Reports, 7, 8710-8710 (2017)
Tensin3 is a novel thyroid-specific gene
Maeda I
Journal of Molecular Endocrinology, 36, R1-R8 (2008)
Epidermal Growth Factor Modulates Tyrosine Phosphorylation of a Novel Tensin Family Member, Tensin311Supported in part by the Shriners Hospitals Research Award and NIH CA102537.
Cui Y
Molecular Cancer Research (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service