Skip to Content
Merck
All Photos(1)

Key Documents

SAB4200190

Sigma-Aldrich

Anti-Sumo-2/3 antibody, Rat monoclonal

clone 3H12, purified from hybridoma cell culture

Synonym(s):

Anti-HSMT3, Anti-MGC117191, Anti-SMT3 homolog 2, Anti-SMT3 suppressor of mif two 3 homolog 2, Anti-SMT3B, Anti-SMT3H2, Anti-sentrin 2, Anti-small ubiquitin-like modifier 2, Anti-small ubiquitin-related modifier 2, Anti-ubiquitin-like protein SMT3B, Monoclonal Anti-Sumo-2/3, SUMO-2/3, SUMO2/3

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

3H12, monoclonal

form

buffered aqueous solution

mol wt

~11 kDa

species reactivity

mouse, rat, human

concentration

~1.0 mg/mL

technique(s)

immunocytochemistry: suitable
western blot: 2-4 μg/mL using HeLa cell nuclear extract

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SUMO3(6612)
mouse ... Sumo3(20610)
rat ... Sumo3(499417)

General description

Monoclonal Anti-Sumo-2/3 (rat IgG2a isotype) is derived from the hybridoma 3H12 produced by the fusion of mouse myeloma cells and splenocytes from rat immunized with a human SUMO3 fusion protein.
Small ubiquitin-like modifier 3 (SUMO3) is encoded by the gene mapped to human chromosome 21q22.3. The protein belongs to the ubiquitin-like protein family and is expressed mainly in cytoplasm.

Immunogen

human SUMO3 fusion protein

Application

Monoclonal Anti-Sumo-2/3 has been used in immunoblotting and immunocytochemistry.

Biochem/physiol Actions

Small ubiquitin-like modifier (SUMO) was identified as a post-translational protein modifier.
Small ubiquitin-like modifier 3 (SUMO3) plays a vital role in enhancing cellular responses to environmental stress. It acts as a key element of the leukemogenic network in transient myeloproliferative disorder (TMD) of the newborn/acute megakaryoblastic leukaemia (AMKL).

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Molecular features of human ubiquitin-like SUMO genes and their encoded proteins
Su HL and Li SS
Gene, 296, 65-73 (2002)
SUMO-2/3 conjugates accumulating under heat shock or MG132 treatment result largely from new protein synthesis.
Castoralova M
Biochimica et Biophysica Acta, 1823, 911-919 (2012)
Functional heterogeneity of small ubiquitin-related protein modifiers SUMO-1 versus SUMO-2/3.
Saitoh H and Hinchey J.
The Journal of Biological Chemistry, 275, 6252-6258 (2000)
The Small Ubiquitin-Like Modifier (SUMO) and SUMO-Conjugating System of Chlamydomonas reinhardtii
Wang Y, et al.
Genetics, 179(1), 177-177 (2008)
A 15q24 microdeletion in transient myeloproliferative disease (TMD) and acute megakaryoblastic leukaemia (AMKL) implicates PML and SUMO3 in the leukaemogenesis of TMD/AMKL
Haemmerling S
British Journal of Haematology, 157, 180-187 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service